Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502139

Sigma-Aldrich

Anti-PDGFR α antibody produced in rabbit

affinity isolated antibody

Synonym(s):

α platelet-derived growth factor receptor precursor, CD140a, PDGF-R-α, PDGFR-α, kinase PDGFR-α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 122 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PGFRA(5156)

General description

Anti-PDGFR Anti-PDGFR α Antibody detects endogenous levels of total PDGFR α protein. Antibody detects endogenous levels of total PDGFR α protein.
Platelet-derived growth factor receptor α (PDGFRA), also known as cluster of differentiation 140a (CD140a), is encoded by the gene mapped to human chromosome 4q12. The encoded protein belongs to the receptor tyrosine kinase gene family.

Immunogen

The antiserum was produced against synthesized peptide derived from human PDGFR alpha.

Immunogen Range: 731-780

Biochem/physiol Actions

Platelet-derived growth factor receptor plays a vital role in the development and maturation of platelets. Platelet-derived growth factor receptor α (PDGFRA) acts as a potential target for imatinib, a revolutionary drug for the treatment of chronic myeloid leukemia. PDGFRA pathway might be involved in developmental process of thrombocytes. The encoded protein facilitates hepatic stellate cell (HSCs) proliferation and migration. Mutation in the gene has been observed in gastrointestinal stromal tumors (GISTs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PDGFR? promoter polymorphisms and expression patterns influence risk of development of imatinib-induced thrombocytopenia in chronic myeloid leukemia: A study from India
Guru S A, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 39(10) (2017)
PDGFRA Mutations in Gastrointestinal Stromal Tumors: Frequency, Spectrum and In Vitro Sensitivity to Imatinib
Corless C L, et al.
Journal of Clinical Oncology, 23(23), 5357-5364 (2005)
A 1.8-Mb YAC contig spanning three members of the receptor tyrosine kinase gene family (Pdgfra, Kit, and Flk1) on mouse chromosome 5.
Brunkow M E, et al.
Genomics, 25(2), 421-432 (1995)
Platelet-Derived Growth Factor Receptor ? Contributes to Human Hepatic Stellate Cell Proliferation and Migration.
Kikuchi A, et al.
The American Journal of Pathology, 187(10), 2273-2287 (2017)
Overexpressed Skp2 within 5p amplification detected by array?based comparative genomic hybridization is associated with poor prognosis of glioblastomas
Saigusa K, et al.
Cancer Science, 96(10), 676-683 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service