Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502021

Sigma-Aldrich

Anti-NOTCH2 (Cleaved-Ala1734) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Motch B, NOTC2, NOTCH2, NTC2, Neurogenic locus notch homolog protein 2 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
WB
clone:
polyclonal
species reactivity:
rat, mouse, human
citations:
4
technique(s):
ELISA: 1:10000
western blot: 1:500-1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 79 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Ala1734)

Gene Information

human ... NOTCH2(4853)

Related Categories

General description

The NOTCH2 (notch 2) gene is mapped to human chromosome 1p12. Anti-NOTCH2 (cleaved-Ala1734) antibody detects endogenous levels of fragment of activated NOTCH2 (cleaved-Ala1734) protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human NOTCH2.

Immunogen Range: 1715-1764

Biochem/physiol Actions

The NOTCH2 (notch2) gene is known to be involved in the proliferation and cell cycle progression in cancer cells. Upregulation of the gene is observed in salivary adenoid cystic carcinoma, and hepatocellular carcinoma. Deletion of NOTCH2 offers tumor cells resistance to chemotherapeutic agents such as 5-fluorouracil. NOTCH2 plays an important role in the regulation of liver development. The NOTCH2 gene is associated with the development of diabetic nephropathy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NOTCH2 mutations cause Alagille syndrome, a heterogeneous disorder of the notch signaling pathway
McDaniell R, et al.
American Journal of Human Genetics, 79(1), 169-173 (2006)
Notch2 is a crucial regulator of self-renewal and tumorigenicity in human hepatocellular carcinoma cells
Wu WR, et al.
Oncology Reports, 36(1), 181-188 (2016)
Altered expression of WFS1 and NOTCH2 genes associated with diabetic nephropathy in T2DM patients
Sharaf SA, et al.
Diabetes Research and Clinical Practice, 140, 304-313 (2018)
Jing Qu et al.
Molecular and cellular biochemistry, 411(1-2), 135-141 (2015-10-03)
Many studies have explored whether the Notch signaling pathway has a tumor-suppressive or an oncogenic role in various tumors; however, the role of the Notch signaling pathway in salivary adenoid cystic carcinoma (SACC) is still unknown. In this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service