Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500465

Sigma-Aldrich

Anti-CRHR1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CRF-R, CRF1, CRH-R 1, Corticotropin-releasing factor receptor 1, Corticotropin-releasing hormone receptor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRHR1(1394)

General description

Anti-CRHR1 Antibody detects endogenous levels of total CRHR1 protein.
CRHR1 (corticotropin releasing factor receptor 1) is a member of the G-protein-coupled receptor superfamily of brain-gut neuropeptides. It is highly expressed in the amygdala, hippocampus and frontal cortex. It has 13 exons and located on human chromosome 17q12-q22.

Immunogen

The antiserum was produced against synthesized peptide derived from human CRHR1.

Immunogen Range: 75-124

Application

Anti-CRHR1 antibody has been used in immunoprecipitation and western blotting.

Biochem/physiol Actions

CRHR1 (corticotropin releasing factor receptor 1) blocks the multiplication of endocrine-related cancer cell lines. It helps to initiate signal transduction pathways, that controls various functions like stress, reproduction, immune response and obesity.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuting Li et al.
Neural plasticity, 2016, 5249893-5249893 (2016-03-01)
Several types of microRNA (miRNA) overexpression in the brain are associated with stress. One of the targets of miR-34c is the stress-related corticotrophin releasing factor receptor 1 mRNA (CRFR1 mRNA). Here we will probe into the short-term effect and long-term
Urocortin protects chondrocytes from NO-induced apoptosis: a future therapy for osteoarthritis?
Intekhab-Alam NY, et al.
Cell Death & Disease (2013)
Dynamic Alterations of miR-34c Expression in the Hypothalamus of Male Rats after Early Adolescent Traumatic Stress
Li C, et al.
Neural Plast. (2016)
N Y Intekhab-Alam et al.
Cell death & disease, 4, e717-e717 (2013-07-13)
Osteoarthritis (OA) is characterized by a loss of joint mobility and pain resulting from progressive destruction and loss of articular cartilage secondary to chondrocyte death and/ or senescence. Certain stimuli including nitric oxide (NO) and the pro-inflammatory cytokine tumor necrosis
Association analysis of polymorphisms of the CRHR1 gene with infantile spasms
Yang G, et al.
Molecular Medicine Reports, 12(2), 2539-2546 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service