Skip to Content
Merck
All Photos(2)

Documents

SAB4200736

Sigma-Aldrich

Anti-S100-A10 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-ANX2LG, Anti-CAL1L, Anti-CLP11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~11 kDa

species reactivity

human

technique(s)

immunoblotting: 1:500-1:1000 using human epidermoid carcinoma A431 cell line extract
immunohistochemistry: 1:500-1:1000 using heat-retrieved formalin-fixed, paraffin-embedded Human Pancrease carcinoma sections

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S100A10(6281)

General description

S100 calcium binding protein A10 (S100-A10), also known as calpactin I light chain or p11 or cellular ligand of annexin II, is a member of the S100 family of proteins. S100-A10 is mapped to human chromosome 1q21.3 and is expressed in the intestine, lungs and kidney. It contains two EF-hand calcium-binding motifs.

Specificity

Anti-S100-A10 recognizes human S100-A10.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminus of human S100-A10, conjugated to KLH

Application

Anti-S100-A10 antibody produced in rabbit may be used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

S100 calcium binding protein A10 (S100-A10) functions as the regulatory subunit of Annexin II (ANX2) to generate the active heterotetramer ANX2/p11 (ANX2T). S100-A10 is implicated in prostate cancer and multiple myeloma and enables cells to reach adhesion and growth at the bone marrow microenvironment. Elevated levels of S100-A10 are observed in squamous non-cell lung cancer, anaplastic large cell lymphoma, renal cell carcinoma and pediatric intracranial ependymoma. Experimental evidence implicates S100-A10 involvement in the mechanism of action of antidepressant drugs and electroconvulsive seizures due to its interaction with specific serotonin receptors.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A role for p11 in the antidepressant action of brain-derived neurotrophic factor
Warner-Schmidt JL, et al.
Biological Psychiatry, 68(6), 528-535 (2010)
Disruption of annexin II/p11 interaction suppresses leukemia cell binding, homing and engraftment, and sensitizes the leukemia cells to chemotherapy
Gopalakrishnapillai A, et al.
Testing, 10(10), e0140564-e0140564 (2015)
Multiple functions of S100A10, an important cancer promoter
Saiki Y and Horii A
Pathology International, 69(11), 629-636 (2019)
p11 and its role in depression and therapeutic responses to antidepressants
Svenningsson P, et al.
Nature Reviews. Neuroscience, 14(10), 673-680 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service