Skip to Content
Merck
All Photos(1)

Documents

SAB4200668

Sigma-Aldrich

Anti-Chromogranin A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CGA, CHGA, chromogranin A (parathyroid secretory protein 1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunohistochemistry: 5-10 μg/mL using heat-retrieved formalin-fixed, paraffin-embedded human pancreas sections

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHGA(1113)

General description

CHGA (chromogranin A) is a granin family acidic glycoprotein. It resides in endocrine and neuroendocrine cells, where it is localized to secretory dense-core granules which store peptide hormones. It is a predominant protein of the chromaffin cell secretory granules.

Immunogen

synthetic peptide corresponding to the GR-44 peptide in the C-terminal region of human Chromogranin-A , conjugated to KLH.

Application

Anti-Chromogranin A antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

CHGA (chromogranin A) is secreted in abnormal levels by neoplastic neuroendocrine cells, which might be linked with tumor growth and progression, as it might influence the different components of tumor stroma. It is a highly sensitive and specific biomarker for the diagnosis of NETs (neuroendocrine tumors). CHGA is processed to produce catestatin (CST), vasostatin-I, serpinin and pancreastatin (PST), which function as metabolic or cardiovascular-regulatory peptides. As this protein is co-stored and co-released with catecholamines, it plays an essential role in production of catecholamine storage vesicles. It functions as a circulating biomarker for gastroenteropancreatic neuroendocrine neoplasm (GEP-NEN) patients.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jibo Hu et al.
Oncology letters, 13(4), 2186-2190 (2017-04-30)
Nonfunctioning pancreatic neuroendocrine carcinoma (NPNEC) is a poorly differentiated neuroendocrine carcinoma and a rare type of pancreatic cancer. Computed tomography (CT)-based imaging is currently the most effective method for diagnosing and classifying NPNEC. The present study investigated the clinical features
Diagnostic value of circulating chromogranin a for neuroendocrine tumors: a systematic review and meta-analysis.
Yang X et al
PLoS ONE, 10(4), e0124884-e0124884 (2015)
Chromogranin A as a marker for diagnosis, treatment, and survival in patients with gastroenteropancreatic neuroendocrine neoplasm.
Wang YH et al
Medicine, 93(27), e247-e247 (2014)
A comparison of three chromogranin A assays in patients with neuroendocrine tumours.
Hoej LB et al
Journal of Gastrointestinal and Liver Diseases : JGLD, 23(4), 419-424 (2014)
A common genetic variant of the chromogranin A-derived peptide catestatin is associated with atherogenesis and hypertension in a Japanese population.
Choi Y et al
Endocrine Journal, 62(9), 797-804 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service