Skip to Content
Merck
All Photos(5)

Documents

SAB4200493

Sigma-Aldrich

Anti-IAPP antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Amylin, Anti-DAP, Anti-Diabetes-associated peptide, Anti-IAP, Anti-Islet amyloid polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat

packaging

antibody small pack of 25 μL

enhanced validation

independent ( Antibodies)
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 5-10 μg/mL using formalin-fixed paraffin-embedded human pancreas.
indirect immunofluorescence: 1-2 μg/mL using rat insulinoma RIN-5F cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Iapp(15874)
rat ... Iapp(24476)

General description

Islet amyloid polypeptide (IAPP, amylin), is a 37-residue peptide hormone co-secreted with insulin from the pancreatic β-cells. IAPP is processed from an 89-amino acid precursor pro-islet amyloid polypeptide (proIAPP) in pancreatic β-cells that undergoes complex posttranslational modifications including protease cleavage, C-terminal amidation and formation of an intramolecular disulfide bridge to produce the mature IAPP.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminus of rat pro-IAPP, conjugated to KLH. The corresponding sequence is identical in mouse IAPP and has 74% sequence identity with human IAPP.

Application

Anti-IAPP antibody produced in rabbit has been used in immunohistochemistry and immunofluorescence.

Biochem/physiol Actions

IAPP/Amylin plays a role in glycemic regulation by slowing gastric emptying and promoting satiety, thereby preventing post-prandial spikes in blood glucose levels. IAPP has been linked to type 2 diabetes (T2DM) and the loss of islet b-cells. The human IAPP (20-29) region that has been found to be essential to amyloid formation is thought to be a key factor in the initiation of amyloid aggregation. Islet amyloid formation, initiated by the aggregation of IAPP, may contribute to this progressive loss of islet β-cells. Amyloid deposits deriving from IAPP are commonly found in pancreatic islets of patients suffering of T2DM, or containing an insulinoma cancer. IAPP, like β-amyloid peptide associated with Alzheimer′s disease, can induce apoptosis in insulin producing β-cells.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service