Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200452

Sigma-Aldrich

Anti-ADFP / PLIN2 antibody, Mouse monoclonal

clone ADFP-5, purified from hybridoma cell culture

Synonym(s):

Adipophilin, Plin2 Antibody, Plin2 Antibody - Monoclonal Anti-ADFP / PLIN2 antibody produced in mouse, Anti-ADRP, Anti-Perilipin 2, Monoclonal Anti-ADFP / PLIN2 antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ADFP-5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~48 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using human HepG2 or SH-SY5Y cells
western blot: 2-4 μg/mL using whole extracts of HEK-293T cells overexpressing human ADFP

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLIN2(123)

General description

Monoclonal Anti-ADFP/PLIN2 (mouse IgG1 isotype) is derived from the hybridoma ADFP-5 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide.
Perilipin 2 (PLIN2) is a 50 kDa major protein belonging to perilipin family. It is present on lipid droplets found in the cytoplasm of variety of cells as hepatocytes, adipocytes, muscle cells, mammary epithelial cells, fibroblasts, endothelial cells and macrophages. PLIN2 gene, spanning 14 kb, with eight exons, is located on human chromosome 9p22.1.

Specificity

Monoclonal Anti-ADFP/PLIN2 recognizes human ADFP/PLIN2.

Immunogen

a synthetic peptide corresponding to a sequence at the C-terminus of human PLIN2, conjugated to KLH.

Application

Monoclonal Anti-ADFP / PLIN2 antibody produced in mouse may be used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

PLIN2 may serve as a biomarker of lipid accumulation in diverse cell types and diseases.
Perilipin 2 (PLIN2) plays a vital role in uptake and storage of lipids as triacylglycerols in lipid droplets in skeletal muscle. PLIN2 is also involved in the regulation of lipid metabolism. High levels of PLIN2 is beneficial for the prevention of insulin resistance. Overexpression of PLIN2 is associated with clear cell renal cell carcinoma (CC-RCC).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20oC in working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivier Sorg et al.
JID innovations : skin science from molecules to population health, 1(4), 100057-100057 (2021-12-16)
In adipocytes and sebocytes, lipid droplet proteins control the storage of lipids in organized droplets and their release on demand. The contribution of lipid droplet proteins to the pathogenesis of acne is plausible because they control the levels of comedogenic
Adipophilin increases triglyceride storage in human macrophages by stimulation of biosynthesis and inhibition of beta-oxidation
Larigauderie G, et al.
FEBS Journal, 273(15), 3498-3510 (2006)
Identification and validation of a 44-gene expression signature for the classification of renal cell carcinomas
Wang Q, et al.
Journal of Experimental & Clinical Cancer Research, 36(1), 176-176 (2017)
Overexpression of PLIN2 is a prognostic marker and attenuates tumor progression in clear cell renal cell carcinoma
Cao Q, et al.
International Journal of Oncology, 53(1), 137-147 (2018)
Perilipin 2 improves insulin sensitivity in skeletal muscle despite elevated intramuscular lipid levels
Bosma M, et al.
Diabetes, DB_111402-DB_111402 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service