Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501032

Sigma-Aldrich

Anti-BMP15 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 43 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BMP15(9210)

Immunogen

Antibody was raised against an 18 amino acid peptide near the amino terminus of human BMP15.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501032.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Galloway et al.
Molecular and cellular endocrinology, 191(1), 15-18 (2002-06-05)
BMP15, also known as growth and differentiation factor 9B (GDF9B), is a member of the transforming growth factor beta superfamily (TGFbeta) which in humans, rodents and sheep is expressed exclusively in the oocyte. BMP15 is closely related to GDF9, another
The transforming growth factor-beta family.
J Massagué
Annual review of cell biology, 6, 597-641 (1990-01-01)
M Laitinen et al.
Mechanisms of development, 78(1-2), 135-140 (1998-12-22)
Growth differentiation factor-9 (GDF-9) is a transforming growth factor-b (TGF-b) family member which is expressed in the oocytes in mouse ovaries (McGrath, S.A., Esquela, A.F., Lee, S.J., 1995. Oocyte-specific expression of growth/differentiation factor-9. Mol. Endocrinol. 9, 131-136). GDF-9 is indispensable
J Aaltonen et al.
The Journal of clinical endocrinology and metabolism, 84(8), 2744-2750 (1999-08-12)
Growth differentiation factor 9 (GDF-9) is a transforming growth factor-beta family member that is required for normal folliculogenesis in female mice, but its role as a regulator of human fertility is still unclear. We determined here by in situ hybridization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service