Skip to Content
Merck
All Photos(1)

Key Documents

SAB2502028

Sigma-Aldrich

Anti-SLC6A4 (C-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

5-HTT, 5-HTTLPR, 5HTT, HTT, solute carrier family 6 (neurotransmitter transporter, serotonin), member 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, bovine, human, mouse

technique(s)

ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC6A4(6532)

General description

Solute carrier family 6 member 4 (SLC6A4) is also known as 5-hydroxytryptamine transporter (5-HTT). It is an integral membrane protein, which is mapped to human chromosome 17q11.2. The encoded protein is a member of the SLC6 (solute carrier family 6) gene family and is mainly expressed in brain and blood cells.

Immunogen

Peptide with sequence C-KERIIKSITPETPT, from the C-terminal region of the protein sequence according to NP_001036.1

Biochem/physiol Actions

Solute carrier family 6 member 4 (SLC6A4) catalyzes the transport of neurotransmitter serotonin from the synaptic space into presynaptic neurons. It plays a vital role in mood and anxiety regulation. Mutation in the gene may alter the expression of serotonin (5HT) in the synaptic cleft, which ultimately leads to schizophrenia. Allelic variations in the gene increase the risk of susceptibility to autism and rigid-compulsive behaviors.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allelic heterogeneity at the serotonin transporter locus (SLC6A4) confers susceptibility to autism and rigid-compulsive behaviors
J S Sutcliffe, et al.
American Journal of Human Genetics, 77(2), 265-279 (2005)
Genetic variation in the serotonin transporter promoter region affects serotonin uptake in human blood platelets
B D Greenberg, et al.
American Journal of Medical Genetics, 88(1), 83-87 (1999)
The solute carrier 6 family of transporters
S Broer, et al.
British Journal of Pharmacology, 167(2), 256-278 (2012)
Linkage mapping of serotonin transporter protein gene SLC6A4 on chromosome 17
J Gelernter, et al.
Human Genetics, 95(6), 677-680 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service