Skip to Content
Merck
All Photos(1)

Documents

SAB2500221

Sigma-Aldrich

Anti-CDKN2A (Isoform 1)/P16INK4A antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Cyclin-dependent kinase inhibitor 2A, Anti-HGNC:1787, Anti-p16INK4a

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDKN2A(1029)

General description

Cyclin dependent kinase inhibitor 2A (CDKN2A), also known as P16-INK4A, belongs to the Ink4 (inhibitor of CDK4) family of CDK inhibitors. It is encoded by the gene mapped to human chromosome 9p21 within the INK4a/ alternate reading frame (ARF) locus. Human P16-INK4A is localized in both nucleus and cytoplasm, and its expression increases with ageing in skin and kidney tissues.

Immunogen

Peptide with sequence C-HARIDAAEGPSDIPD from the C Terminus of the protein sequence according to NP_000068.1.

Biochem/physiol Actions

Cyclin dependent kinase inhibitor 2A (CDKN2A), also known as P16-INK4A, arrests cell cycle progression by inhibiting S-phase and promotes apoptosis. The encoded protein has anti-proliferative biological activity, and is implicated in the retinoblastoma (Rb) protein pathway. P16-INK4A acts as a tumor suppressor and plays a vital role in ageing and cellular senescence. Increased expression of this protein might lead to various types of high grade cancers. P16-INK4A also facilitates cell invasion and angiogenesis. Bronchial lavage P16INK4A promoter methylation might function as a potential biomarker for diagnosis of lung cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bronchial lavage P 16INK4A gene promoter methylation and lung cancer diagnosis: A meta-analysis.
Yifan D
Indian Journal of Cancer (2015)
Expression and significance of p53, rb, p21/waf-1, p16/ink-4a, and PTEN tumor suppressors in canine melanoma.
Koenig A
Veterinary Pathology (2002)
p16(Ink4a) overexpression in cancer: a tumor suppressor gene associated with senescence and high-grade tumors.
Romagosa C
Oncogene (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service