Skip to Content
Merck
All Photos(3)

Key Documents

SAB2108482

Sigma-Aldrich

Anti-SNAI1

affinity isolated antibody

Synonym(s):

Anti- SLUGH2, Anti- SNAH, Anti- SNAIL, Anti- SNAIL1, Anti-SNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
guinea pig, human, rat
citations:
4
technique(s):
immunoblotting: suitable
immunohistochemistry: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

29 kDa

species reactivity

guinea pig, human, rat

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

accession no.

NM_005985

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNAI1(6615)

Immunogen

Synthetic peptide directed towards the N terminal region of human SNAI1

Biochem/physiol Actions

The Drosophila embryonic protein snail is a zinc finger transcriptional repressor which downregulates the expression of ectodermal genes within the mesoderm. The nuclear protein encoded by this gene is structurally similar to the Drosophila snail protein, and is also thought to be critical for mesoderm formation in the developing embryo.

Sequence

Synthetic peptide located within the following region: MPRSFLVRKPSDPNRKPNYSELQDSNPEFTFQQPYDQAHLLAAIPPPEIL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulgi Park et al.
Endocrinology, 161(2) (2020-01-03)
SULT2B1b (SULT2B) is a prostate-expressed hydroxysteroid sulfotransferase, which may regulate intracrine androgen homeostasis by mediating 3β-sulfation of dehydroepiandrosterone (DHEA), the precursor for 5α-dihydrotestosterone (DHT) biosynthesis. The aldo-keto reductase (AKR)1C3 regulates androgen receptor (AR) activity in castration-resistant prostate cancer (CRPC) by
Kaori Yoriki et al.
Scientific reports, 9(1), 6697-6697 (2019-05-02)
Estrogen-related receptor alpha (ERRα), which shares structural similarities with estrogen receptors, is associated with tumor progression in endometrial cancer, but little is known about the detailed underlying mechanism. We investigated whether ERRα, in cooperation with peroxisome proliferator-activated receptor gamma coactivator
Tinte Itinteang et al.
Plastic and reconstructive surgery. Global open, 8(2), e2598-e2598 (2020-04-21)
We have previously shown that the endothelium of the microvessels of infantile hemangioma (IH) exhibits a hemogenic endothelium phenotype and proposed its potential to give rise to mesenchymal stem cells, similar to the development of hematopoietic cells. This endothelial-to-mesenchymal transition
Han Bao et al.
Oncology letters, 14(3), 2831-2837 (2017-09-21)
Long non-coding RNAs (lncRNAs) serve an important role in numerous human diseases, including cancer. Abnormal expression of lncRNAs has been associated with a number of tumor types; however, the underlying mechanisms through which lncRNA functions have yet to be elucidated.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service