Skip to Content
Merck
All Photos(3)

Documents

SAB2108476

Sigma-Aldrich

Anti-MYC

affinity isolated antibody

Synonym(s):

Anti- MRTL, Anti- bHLHe39, Anti-c-Myc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

50 kDa

species reactivity

guinea pig, mouse, rabbit, human, rat

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

accession no.

NM_002467

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

The cellular myelocytomatosis (c-myc) gene mapped to human chromosome 8q24, is the cellular homologue of the v-myc gene originally isolated from an avian myelocytomatosis virus. c-myc is a member of MYC gene family. c-Myc gene codes for basic helix-loop-helix/leucine zipper (bHLH/LZ) transcription factor that regulates the G1-S cell cycle transition.

Immunogen

Synthetic peptide directed towards the N terminal region of human MYC

Biochem/physiol Actions

The cellular myelocytomatosis (c-myc) oncogene plays a vital role in cellular proliferation, differentiation, apoptosis and acts as transcriptional regulator of gene expression. c-Myc expression is essential and sufficient to assist most of the cells to enter synthetic (S) phase of the cell cycle. The encoded protein plays a crucial role in vasculogenesis and angiogenesis during cancer development and progression. c-Myc interacts with its binding partner Max and activates the transcription of growth promoting genes such as cyclin D2, ornithine decarboxylase and E2F1 and it also represses the transcription of multiple genes, especially p21 and p27, by binding to the transcription initiator element (Inr) in a complex with Max and either Sp1 or Miz1. Overexpression of MYC in DLBCL (diffuse large B-cell lymphoma) results in poor outcome and invasive treatment when medicated with rituximab plus cyclophosphamide, doxorubicin, vincristine and prednisone (R-CHOP).

Sequence

Synthetic peptide located within the following region: MDFFRVVENQQPPATMPLNVSFTNRNYDLDYDSVQPYFYCDEEENFYQQQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

8q24 prostate, breast, and colon cancer risk loci show tissue-specific long-range interaction with MYC.
Ahmadiyeh, Nasim, et al.
Proceedings of the National Academy of Sciences of the USA, 107, 9742-9746 (2010)
8q24 prostate, breast, and colon cancer risk loci show tissue-specific long-range interaction with MYC
Ahmadiyeh N, et al.
Proceedings of the National Academy of Sciences of the USA, 107, 9742-9746 (2010)
Xiaoyong Chen et al.
Virologica Sinica, 36(5), 1027-1035 (2021-04-09)
Host interferon-stimulated gene 20 (ISG20) exerts antiviral effects on viruses by degrading viral RNA or by enhancing IFN signaling. Here, we examined the role of ISG20 during pseudorabies virus (PRV) proliferation. We found that ISG20 modulates PRV replication by enhancing
Apoptotic signaling by c-MYC.
Hoffman B and Liebermann DA.
Oncogene, 27, 6462-6472 (2008)
Troy A Baudino et al.
Genes & development, 16(19), 2530-2543 (2002-10-09)
c-Myc promotes cell growth and transformation by ill-defined mechanisms. c-myc(-/-) mice die by embryonic day 10.5 (E10.5) with defects in growth and in cardiac and neural development. Here we report that the lethality of c-myc(-/-) embryos is also associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service