Skip to Content
Merck
All Photos(1)

Key Documents

SAB2104288

Sigma-Aldrich

Anti-THPO antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC163194, Anti-MGDF, Anti-MKCSF, Anti-ML, Anti-MPLLG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... THPO(7066)

General description

Thrombopoietin (TPO) is a lineage specific growth factor, produced in the liver, kidney and skeletal muscle. It is an acidic glycoprotein. The gene encoding it is localized on human chromosome 3q27.1.

Immunogen

Synthetic peptide directed towards the middle region of human THPO

Biochem/physiol Actions

Thrombopoietin (TPO) stimulates the proliferation and maturation of megakaryocytes, and promotes increased circulating levels of platelets in vivo. TPO signals through the cellular proto-oncogene (c-mpl) receptor and acts as an important regulator of circulating platelets. High levels of TPO have been observed in patients with aplastic anemia.

Sequence

Synthetic peptide located within the following region: NLQPGYSPSPTHPPTGQYTLFPLPPTLPTPVVQLHPLLPDPSAPTPTPTS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The molecular mechanisms that control thrombopoiesis.
Kaushansky K
The Journal of Clinical Investigation, 115(12), 3339-3347 (2005)
Majed J Dasouki et al.
Blood, 122(20), 3440-3449 (2013-10-03)
We recently identified 2 siblings afflicted with idiopathic, autosomal recessive aplastic anemia. Whole-exome sequencing identified a novel homozygous missense mutation in thrombopoietin (THPO, c.112C>T) in both affected siblings. This mutation encodes an arginine to cysteine substitution at residue 38 or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service