Skip to Content
Merck
All Photos(1)

Key Documents

SAB2102177

Sigma-Aldrich

Anti-SLC22A6 (ab2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HOAT1, Anti-MGC45260, Anti-OAT1, Anti-PAHT, Anti-Solute carrier family 22 (organic anion transporter), member 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

62 kDa

species reactivity

mouse, guinea pig, human, horse, rat, bovine, rabbit, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC22A6(9356)

General description

Solute carrier family 22 member 6 (SLC22A6) belongs to organic anion transporter-1 family. The protein is expressed in kidney and brain. The gene is located on human chromosome 11q12.3.

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC22A6

Biochem/physiol Actions

Solute carrier family 22 member 6 (SLC22A6) is involved in the sodium-dependent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and may be localized to the basolateral membrane. Four transcript variants encoding four different isoforms have been found for this gene.
SLC22A6 regulates the body disposition of a variety of clinically important drugs, such as, anti-HIV therapeutics, antitumor drugs, antibiotics, antihypertensives and anti-inflammatories. Ubiquitination sites of SLC22A6 is involved in the protein kinase C (PKC)-mediated endocytosis of the transporter. It plays an important role in substrate recognition.

Sequence

Synthetic peptide located within the following region: AFNDLLQQVGGVGRFQQIQVTLVVLPLLLMASHNTLQNFTAAIPTHHCRP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Three ubiquitination sites of organic anion transporter-1 synergistically mediate protein kinase C-dependent endocytosis of the transporter
Li S, et al.
Molecular Pharmacology, mol-113 (2013)
Organic anion transporter OAT1 undergoes constitutive and protein kinase C-regulated trafficking through a dynamin-and clathrin-dependent pathway
Zhang Q, et al.
The Journal of Biological Chemistry (2008)
Navaz Karimian Pour et al.
Pharmaceutics, 11(12) (2019-11-27)
Inflammation impacts the expression and function of drug transporters at term-gestation; however, the impact of inflammation on the expression of drug transporters at mid-gestation is largely unknown. Since renal drug transporters play a key role in the clearance of many
Functional role of the C terminus of human organic anion transporter hOAT1
Xu W, etal.
The Journal of Biological Chemistry, 281(42), 31178-31183 (2006)
Josefin A Jacobsson et al.
Genomics, 90(5), 595-609 (2007-08-24)
The solute carrier family 22 (SLC22) is a large family of organic cation and anion transporters. These are transmembrane proteins expressed predominantly in kidneys and liver and mediate the uptake and excretion of environmental toxins, endogenous substances, and drugs from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service