Skip to Content
Merck
All Photos(2)

Key Documents

SAB1409948

Sigma-Aldrich

Monoclonal Anti-CDKL1 antibody produced in mouse

clone 8B2, ascites fluid, solution

Synonym(s):

KKIALRE, p42

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
WB
clone:
8B2, monoclonal
species reactivity:
human
citations:
technique(s):
indirect ELISA: suitable
western blot: 1:500-1:1000

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

8B2, monoclonal

form

solution

mol wt

antigen 36.63 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDKL1(8814)

General description

This gene product is a member of a large family of CDC2-related serine/threonine protein kinases. It accumulates primarily in the nucleus. Alternative splice variants encoding different protein isoforms have been described, but their full-length nature has not been determined. (provided by RefSeq)

Immunogen

CDKL1 (NP_004187, 259 a.a. ~ 358 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
YPALGLLKGCLHMDPTQRLTCEQLLHHPYFENIREIEDLAKEHNKPTRKTLRKSRKHHCFTETSKLQYLPQLTGSSILPALDNKKYYCDTKKLNYRFPNI

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service