Skip to Content
Merck
All Photos(4)

Key Documents

SAB1405936

Sigma-Aldrich

Anti-HLA-A antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~40.8 kDa

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HLA-A(3105)

Related Categories

General description

HLA-A belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-A alleles have been described. (provided by RefSeq)

The previously assigned protein identifier P01891 has been merged into P04439. Full details can be found on the UniProt database.

Immunogen

HLA-A (NP_002107.3, 1 a.a. ~ 365 a.a) full-length human protein.

Sequence
MAVMAPRTLLLLLSGALALTQTWAGSHSMRYFFTSVSRPGRGEPRFIAVGYVDDTQFVRFDSDAASQRMEPRAPWIEQEGPEYWDQETRNVKAQSQTDRVDLGTLRGYYNQSEAGSHTIQIMYGCDVGSDGRFLRGYRQDAYDGKDYIALNEDLRSWTAADMAAQITKRKWEAAHEAEQLRAYLDGTCVEWLRRYLENGKETLQRTDPPKTHMTHHPISDHEATLRCWALGFYPAEITLTWQRDGEDQTQDTELVETRPAGDGTFQKWAAVVVPSGEEQRYTCHVQHEGLPKPLTLRWELSSQPTIPIVGIIAGLVLLGAVITGAVVAAVMWRRKSSDRKGGSYTQAASSDSAQGSDVSLTACKV

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amal Ghraieb et al.
Journal of autoimmunity, 91, 61-72 (2018-04-24)
Alopecia areata (AA) is understood to be a CD8+/NKG2D+ T cell-dependent autoimmune disease. Here, we demonstrate that human AA pathogenesis of is also affected by iNKT10 cells, an unconventional T cell subtype whose number is significantly increased in AA compared to healthy
Sevim Barbasso Helmers et al.
Rheumatology (Oxford, England), 57(12), 2149-2157 (2018-08-14)
To investigate the expression of IL-18 in symptomatic and asymptomatic muscle tissues of patients with PM and DM and the effects of conventional immunosuppressive treatment on such expression. Two cohorts of patients were included in this study. The first cohort
Carmen Lefaucheur et al.
Kidney international, 96(1), 189-201 (2019-04-22)
Anti-angiotensin II type 1 receptor (AT1R) antibodies have been associated with allograft rejection. We hypothesized that circulating AT1R antibodies might identify kidney transplant recipients at increased risk of allograft rejection and loss who are not identified by the HLA system.
Natalia F Smirnova et al.
JCI insight, 4(3) (2019-02-08)
Lung transplantation (LTx) is the only therapeutic option for many patients with chronic lung disease. However, long-term survival after LTx is severely compromised by chronic rejection (chronic lung allograft dysfunction [CLAD]), which affects 50% of recipients after 5 years. The
Olivier Aubert et al.
Journal of the American Society of Nephrology : JASN, 30(4), 625-639 (2019-03-16)
Transplant glomerulopathy, a common glomerular lesion observed after kidney transplant that is associated with poor prognosis, is not a specific entity but rather the end stage of overlapping disease pathways. Its heterogeneity has not been precisely characterized to date. Our

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service