Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403761

Sigma-Aldrich

Monoclonal Anti-EIF4EBP1 antibody produced in mouse

clone 1F7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

4E-BP1, 4EBP1, BP-1, MGC4316, PHAS-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~39.09 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EIF4EBP1(1978)

General description

This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E (eIF4E), which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5′ end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation. (provided by RefSeq)

Immunogen

EIF4EBP1 (AAH04459, 1 a.a. ~ 118 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSGGSSCSQTPSRAIPATRRVVLGDGVQLPPGDYSTTPGGTLFSTTPGGTRIIYDRKFLMECRNSPVTKTPPRDLPTIPGVTSPSSDEPPMEASQSHLRNSPEDKRAGGEESQFEMDI

Biochem/physiol Actions

The gene EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1) encodes a translation repressor that interacts with eukaryotic translation initiation factor 4E (eIF4E). eIF4E is a multisubunit complex that recruits 40S ribosomal subunits to the 5′ end of mRNA. The interaction of the encoded binding protein with this complex inhibits translation. The phosphorylation of eIF4Ebp1 in response to stimuli such as, UV irradiation and insulin signaling, results in dissociation of this factor from the eIF4E complex, leading to initiation of translation of mRNA. The encoded protein participates in cell proliferation, apoptosis, invasion, and metastasis. It functions as an effector molecule in mTOR (mammalian target of rapamycin complex 1) signaling pathway that regulates protein synthesis. It is found to be overexpressed in hepatocellular carcinoma tissues and serves as a potential prognostic and therapeutic target.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of 4E-BP1 phosphorylation: a novel two-step mechanism.
Gingras AC, et al.
Genes & Development, 13, 1422-1437 (1999)
eIF4E binding protein 1 expression is associated with clinical survival outcomes in colorectal cancer.
Chao MW, et al.
Oncotarget, 6(27), 24092-24104 (2015)
EIF4EBP1 overexpression is associated with poor survival and disease progression in patients with hepatocellular carcinoma.
Cha YL, et al.
PLoS ONE, 10(2) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service