Skip to Content
Merck
All Photos(2)

Documents

SAB1402301

Sigma-Aldrich

Monoclonal Anti-PCP4 antibody produced in mouse

clone 1E3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

PEP-19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~32.93 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCP4(5121)

Related Categories

General description

Purkinje cell protein 4 (PCP4) is an anti-apoptotic, calmodulin-binding peptide which is expressed in neural cells. It is also expressed in the Purkinje cells of the cerebellum, kidneys, prostate and the uterus. PCP4 is a 7.6kDa with an IQ-motif. The gene encoding this protein is localized on human chromosome 21q22.2.

Immunogen

PCP4 (NP_006189.2, 1 a.a. ~ 62 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSERQGAGATNGKDKTSGENDGQKKVQEEFDIDMDAPETERAAVAIQSQFRKFQKKKAGSQS

Biochem/physiol Actions

Purkinje cell protein 4 (PCP4) may have a role in apoptosis and cellular degeneration. It acts as an accelerator of calcium association and disassociation with calmodulin. The protein also functions in synaptic plasticity.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PCP4 maps between D21S345 and P31P10SP6 on chromosome 21q22.2-->q22.3.
Hubert RS and Korenberg JR
Cytogenetics and Cell Genetics (1997)
Anti-apoptotic effects of PCP4/PEP19 in human breast cancer cell lines: a novel oncotarget.
Hamada T
Oncotarget (2014)
PCP4: a regulator of aldosterone synthesis in human adrenocortical tissues.
Journal of Molecular Endocrinology (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service