Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
human
Quality Segment
recombinant
expressed in HEK 293 cells
assay
≥95% (SDS-GE)
form
lyophilized powder
mol wt
32 kDa by SDS-PAGE (nonglycosylated form, under reducing conditions), 35 kDa by SDS-PAGE (glycosylated form, under reducing conditions)
technique(s)
ligand binding assay: suitable
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... FASLG(356)
Application
Biochem/physiol Actions
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| recombinant expressed in HEK 293 cells | recombinant expressed in CHO cells | recombinant expressed in HEK 293 cells | recombinant expressed in HEK 293 cells |
| assay ≥95% (SDS-GE) | assay >95% (SDS-PAGE) | assay >95% (SDS-PAGE) | assay >90% (SDS-PAGE) |
| biological source human | biological source - | biological source mouse | biological source mouse |
| technique(s) ligand binding assay: suitable | technique(s) - | technique(s) - | technique(s) - |
| mol wt 32 kDa by SDS-PAGE (nonglycosylated form, under reducing conditions), 35 kDa by SDS-PAGE (glycosylated form, under reducing conditions) | mol wt monomer calculated mol wt ~18 kDa, 26-28 kDa by SDS-PAGE | mol wt ~34 kDa by SDS-PAGE | mol wt ~22 kDa by SDS-PAGE |
| form lyophilized powder | form lyophilized powder | form lyophilized | form liquid |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



