Skip to Content
Merck
All Photos(1)

Documents

S7947

Sigma-Aldrich

Stattic

≥98% (HPLC), powder

Synonym(s):

6-Nitrobenzo[b]thiophene-1,1-dioxide, Stat three inhibitory compound

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5NO4S
CAS Number:
Molecular Weight:
211.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >20 mg/mL

SMILES string

[O-][N+](=O)c1ccc2C=CS(=O)(=O)c2c1

InChI

1S/C8H5NO4S/c10-9(11)7-2-1-6-3-4-14(12,13)8(6)5-7/h1-5H

InChI key

ZRRGOUHITGRLBA-UHFFFAOYSA-N

Application

Stattic has been used to inhibit signal transducer and activator of transcription 3 (STAT3) activity in various cell lines and culture.
Stattic was used to study Stat3-mediated cell signaling in human lung carcinoma cells.5

Biochem/physiol Actions

Stattic (Stat3 three inhibitory compound) alters the SH2 domain of Stat3 and indirectly inhibits with phosphopeptide binding. It is readily transported across the cell membrane compared to other phosphopeptides.2 Stattic induces increased formation of ROS and negatively affects the cardiomyocyte mitochondrial function,3 and sensitizes nasopharyngeal carcinoma cells to chemoradiotherapy.4
Stattic is an irreversible STAT3 activation inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long noncoding RNA PVT1 promotes hepatoblastoma cell proliferation through activating STAT3
Luo Z and Cao P
Cancer Management and Research, 11, 8517-8517 (2019)
Human dental pulp stem cells grown in neurogenic media differentiate into endothelial cells and promote neovasculogenesis in the mouse brain.
Luzuriaga J, et al.
Frontiers in Physiology, 10, 347-347 (2019)
Yunbao Pan et al.
PloS one, 8(1), e54565-e54565 (2013-02-06)
Nasopharyngeal carcinoma (NPC) is an Epstein-Barr virus-associated malignancy most common in East Asia, Africa and Alaska. Radiotherapy and cisplatin-based chemotherapy are the main treatment options. Unfortunately, disease response to concurrent chemoradiotherapy varies among patients with NPC, and many cases are
Nikolaos Sismanopoulos et al.
PloS one, 7(3), e33271-e33271 (2012-03-14)
Interleukin 9 (IL-9) has been implicated in mast cell-related inflammatory diseases, such as asthma, where vascular endothelial growth factor (VEGF) is involved. Here we report that IL-9 (10-20 ng/ml) induces gene expression and secretion of VEGF from human LAD2. IL-9
Proatherogenic stimuli induce HuR in atherosclerosis through MAPK/ErK pathway
Cheng M, et al.
American Journal of Translational Research, 11(4), 2317-2317 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service