Skip to Content
Merck
All Photos(1)

Documents

S7076

Sigma-Aldrich

SB 202190 monohydrochloride hydrate

≥98% (HPLC), solid, p38 MAP kinase inhibitor

Synonym(s):

4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)-1H-imidazole monohydrochloride hydrate, 4-[4-(4-Fluorophenyl)-5-(4-pyridinyl)-1H-imidazol-2-yl]phenol monohydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14FN3O · HCl · xH2O
CAS Number:
Molecular Weight:
367.80 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

SB 202190 monohydrochloride hydrate, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to beige

solubility

DMSO: ≥12 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

O.Cl.Oc1ccc(cc1)-c2nc(-c3ccc(F)cc3)c([nH]2)-c4ccncc4

InChI

1S/C20H14FN3O.ClH.H2O/c21-16-5-1-13(2-6-16)18-19(14-9-11-22-12-10-14)24-20(23-18)15-3-7-17(25)8-4-15;;/h1-12,25H,(H,23,24);1H;1H2

InChI key

CZZOICWZVYZUNP-UHFFFAOYSA-N

Application

SB 202190 may be used to inhibit p38-mediated cell signaling.
SB 202190 monohydrochloride hydrate has been used as a component of human prostate culture medium and organoid culture medium It has also been used as p38 mitogen activated protein kinase (MAPK) inhibitor in colon cancer cells.

Biochem/physiol Actions

SB 202190 binds within the ATP pocket of p38 kinase and inhibits the p38α and β isoforms.
SB 202190 monohydrochloride hydrate mediates the phosphorylation of serine in the mutant p53 protein and modulates cell adhesion and prevents cell proliferation in breast cancer cell lines.
SB 202190 is a highly selective, potent, and cell-permeable inhibitor of p38 MAP kinase.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoid culture of human prostate cancer cell lines LNCaP and C4-2B
Ma L, et al.
American journal of clinical and experimental urology, 5(3), 25-25 (2017)
Organoid culture systems for prostate epithelial and cancer tissue
Drost J, et al.
Nature Protocols, 11(2), 347-347 (2016)
Differential effects of p38 MAP kinase inhibitors SB203580 and SB202190 on growth and migration of human MDA-MB-231 cancer cell line
Duzgun SA, et al.
Cytotechnology, 69(4), 711-724 (2017)
Hanbing Song et al.
Nature communications, 13(1), 141-141 (2022-01-12)
Prostate cancer is the second most common malignancy in men worldwide and consists of a mixture of tumor and non-tumor cell types. To characterize the prostate cancer tumor microenvironment, we perform single-cell RNA-sequencing on prostate biopsies, prostatectomy specimens, and patient-derived
Rui Wang et al.
Genome medicine, 14(1), 93-93 (2022-08-17)
Colorectal cancer (CRC) ranks as the second-leading cause of cancer-related death worldwide with metastases being the main cause of cancer-related death. Here, we investigated the genomic and transcriptomic alterations in matching adjacent normal tissues, primary tumors, and metastatic tumors of

Articles

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service