Skip to Content
Merck
All Photos(1)

Key Documents

S5692

Supelco

SYPRO® Orange Protein Gel Stain

Synonym(s):

SYPRO® dye, protein gel stain

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41105322
NACRES:
NA.32
Pricing and availability is not currently available.

shelf life

≥6 mo. (when stored desiccated and protected from light at room temperature, 4 °C or 20 °C.)

technique(s)

protein staining: suitable

fluorescence

λex 300,470 nm; λem 570 nm

General description

SYPRO Orange is similar to SYPRO Red although it is somewhat brighter and gives slightly higher background fluorescence. For those using a laser-excited gel scanner, an argon-ion laser-based instrument is recommended for SYPRO Orange. The dye is efficiently excited by UV or broad band illumination. It is not suitable for staining proteins on blots or in IEF gels and shows reduced sensitivity when staining proteins on 2-D gels.
SYPRO Orange is:
  • Highly sensitive. The stain can detect 1 to 2 ng of protein per minigel band, making it more sensitive than Coomassie Brilliant Blue or silver staining.
  • Rapid. Staining is complete in less than one hour
  • Simple. After electrophoresis, the gel is stained, rinsed and photographed; no fixation or destaining steps are required
  • Compatible with standard laboratory equipment. Stained proteins can be visualized with a standard 300 nm UV transilluminator or laser scanner.
  • Cost-effective. Staining with SYPRO Orange is less expensive than silver staining and requires less time.
  • Low protein-to-protein variability. The dye interacts with the SDS coat around the protein, giving more consistent staining between different types of proteins compared to Coomassie or silver staining.
  • Selective for proteins. SYPRO Orange detects proteins as small as 6.5 kDa and does not stain nucleic acids or lipopolysaccharides. It does stain glycosylated proteins.
  • Broad linear range of detection. The fluorescence intensity of the stained bands is linear with protein quantity over three orders of magnitude (a much broader range than either Coomassie or silver staining).

Application

SYPRO orange protein gel stain has been used in the thermal denaturation assay and thermal shift assay of protein(s) of interest.[1][2][3] It has also been used in differential scanning fluorimetry.[4]

Caution

The SYPRO stock solutions should be stored desiccated and protected from light at room temperature, 4 °C or 20 °C. When stored properly, these stock solutions are stable for six months to one year. The staining reagent diluted in buffer or acetic acid solution can be stored in very clean and detergent-free glass or plastic bottles, protected from light at 4 °C for at least three months. 

Legal Information

SYPRO is a registered trademark of Life Technologies

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiahezi Kuai et al.
Frontiers in plant science, 8, 1715-1715 (2017-10-20)
Each year, crop yield is lost to weeds competing for resources, insect herbivory and diseases caused by pathogens. To thwart these insults and preserve yield security and a high quality of traits, conventional agriculture makes use of improved cultivars combined
Mohammad M Rahman et al.
Journal of bacteriology, 201(20) (2019-07-31)
The gastric pathogen Helicobacter pylori has limited ability to use carbohydrates as a carbon source, relying instead on exogenous amino acids and peptides. Uptake of certain peptides by H. pylori requires an ATP binding cassette (ABC) transporter annotated dipeptide permease
Brijesh B Khadgi et al.
Nucleic acids research, 47(16), 8708-8719 (2019-08-09)
Long Interspersed Elements (LINEs), also known as non-LTR retrotransposons, encode a multifunctional protein that reverse transcribes its mRNA into DNA at the site of insertion by target primed reverse transcription. The second half of the integration reaction remains very poorly
Ruth Cohen-Khait et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(52), 14982-14987 (2016-12-14)
Protein-protein interactions occur via well-defined interfaces on the protein surface. Whereas the location of homologous interfaces is conserved, their composition varies, suggesting that multiple solutions may support high-affinity binding. In this study, we examined the plasticity of the interface of
M Murshida Mahbub et al.
Mobile DNA, 8, 16-16 (2017-11-21)
R2 elements are a clade of early branching Long Interspersed Elements (LINEs). LINEs are retrotransposable elements whose replication can have profound effects on the genomes in which they reside. No crystal or EM structures exist for the reverse transcriptase (RT)

Articles

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

See All

Protocols

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Questions

1–3 of 3 Questions  
  1. サイプロオレンジ s5692 50ul の濃度は5mMですか?

    1 answer
    1. This product is provided as a 5000X concentration in DMSO. The specific concentration of this dye is considered proprietary.

      Helpful?

  2. It's used for Protein quantification assay?

    1 answer
    1. This reagent is suitable for the detection of proteins. However, protein quantitation typically utilizes other methods, as described in the graphic below. Please see the following link to review the product datasheet for additional information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/161/599/s5692dat.pdf

      Helpful?

  3. 1. Is the SYPRO Orange Protein Gel Stain a 5,000x concentrate?
    2. Is there a protocol available for using the SYPRO Orange protein stain for a thermal denaturation assay?

    1 answer
    1. The SYPRO Orange Protein Gel Stain is indeed provided as a 5000X concentrated solution in DMSO.
      Regarding a protocol for using the SYPRO Orange protein stain for a thermal denaturation assay, the product information is focused on gels. While the product is not specifically tested for this application,

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service