Skip to Content
Merck
All Photos(3)

Key Documents

S4072

Sigma-Aldrich

Anti-Sirt3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SIR2L3, Anti-Sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~28 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 2.5-5 μg/mL using human HepG2 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIRT3(23410)

General description

Sirtuin 3 (Sirt3) is an NAD-dependent deacetylase localized to the mitochondrial matrix. SIRT3 encodes a 44 kDa full length form and a N-terminal 142 amino acids truncated form. It is mapped to human chromosome 11p15.5.

Immunogen

synthetic peptide corresponding to amino acids 384-399 of human SIRT3, conjugated to KLH via an N-terminal cysteine residue.

Application

Anti-Sirt3 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Sirtuin 3 (Sirt3) is mainly a nuclear protein that transfers to the mitochondria during cellular stress. In humans, Sirt3 deacetylates and activates the mitochondrial enzyme acetyl-CoA synthetase 1 (AceCS2). Sirt3 is implicated in regulating mitochondrial function and thermogenesis in brown adipocytes. Its levels have been shown to correlate with extended life span. Increased levels of Sirt3 were associated with node positive breast cancer.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of a bidirectional promoter shared between two human genes related to aging: SIRT3 and PSMD13
Bellizzi D, et al.
Genomics, 89(1), 143-150 (2007)
Jing Zhang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(12), 4418-4431 (2014-03-22)
Modern society enables a shortening of sleep times, yet long-term consequences of extended wakefulness on the brain are largely unknown. Essential for optimal alertness, locus ceruleus neurons (LCns) are metabolically active neurons that fire at increased rates across sustained wakefulness.
Komuraiah Myakala et al.
The Journal of biological chemistry, 299(8), 104975-104975 (2023-07-11)
Diabetes mellitus is the leading cause of cardiovascular and renal disease in the United -States. Despite the beneficial interventions available for patients with diabetes, there remains a need for additional therapeutic targets and therapies in diabetic kidney disease (DKD). Inflammation
SIRT3: oncogene and tumor suppressor in cancer
Torrens-Mas M, et al.
Cancers, 9(7), 90-90 (2017)
SirT3 is a nuclear NAD+-dependent histone deacetylase that translocates to the mitochondria upon cellular stress
Scher MB, et al.
Genes & Development, 21(8), 920-928 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service