Skip to Content
Merck
All Photos(1)

Key Documents

S3271

Micro-dissecting scissors

size 4 in., curved, sharp point stainless steel

Synonym(s):

Dissection Scissors, Scissors for Dissection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122408
NACRES:
NB.35
Pricing and availability is not currently available.

material

curved, sharp point stainless steel

size

4 in.

Looking for similar products? Visit Product Comparison Guide

Application

Micro-dissecting scissors have been used to make an incise in the skin and peritoneum during adrenalectomy in mice.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service