Skip to Content
Merck
All Photos(1)

Key Documents

S2816

Sigma-Aldrich

SKF-83959 hydrobromide

≥98% (HPLC), solid

Synonym(s):

6-Chloro-7,8-dihydroxy-3-methyl-1-(3-methylphenyl)-2,3,4,5-tetrahydro-1H-3-benzazepine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20NClO2 · HBr
CAS Number:
Molecular Weight:
398.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >20 mg/mL
H2O: insoluble

SMILES string

Br[H].CN1CCc2c(Cl)c(O)c(O)cc2C(C1)c3cccc(C)c3

InChI

1S/C18H20ClNO2.BrH/c1-11-4-3-5-12(8-11)15-10-20(2)7-6-13-14(15)9-16(21)18(22)17(13)19;/h3-5,8-9,15,21-22H,6-7,10H2,1-2H3;1H

InChI key

FHYWNBUFNGHNCP-UHFFFAOYSA-N

Gene Information

human ... DRD1(1812)

Application

SKF-83959 was used in dopamine signaling studies on eye blinking in monkeys and on phosphorylation of CaMKIIα in mice.

Biochem/physiol Actions

SKF-83959 is an atypical agonist of D1 dopamine receptor and an allosteric modulator of σ-1 receptor on in brain and liver tissues. It has neuroprotective and anti-parkinsonian effects through selective activation of phosphoinositol-linked D1 receptor and promotes migration of cultures astrocytes by ERK1/2 activation. In in vitro studies SKF-83959 exhibits antagonist activity against dopamine D1 receptors coupled to adenylyl cyclase.
D1 dopamine receptor agonist. Displays antagonist activity in vitro and agonist activity in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Fujita et al.
European journal of pharmacology, 632(1-3), 39-44 (2010-02-04)
This study applies new magnetic sensor-electromyographic technology for recording jaw movements in freely moving rats to analyse topographies of orofacial movement that occur in association with individual elements of behaviour under challenge with two dopamine D(1)-like receptor agonists, SKF 83822
Emily M Jutkiewicz et al.
The Journal of pharmacology and experimental therapeutics, 311(3), 1008-1015 (2004-08-05)
Dopamine D1 ligands have been classified and ordered according to efficacy in both in vitro and in vivo studies. In the present experiments, dopamine D1 ligands reported to differ in in vitro efficacy were evaluated for efficacy-related effects on eye
A R Cools et al.
Neuropharmacology, 42(2), 237-245 (2002-01-24)
SKF 83959 that has a unique antiparkinson profile in animal models of Parkinson's disease is an in vitro dopamine D1 antagonist of receptors coupled to adenylyl cyclase. We hypothesized that SKF 83959, among others, interacts with dopamine D1 receptors coupled
Hong-Yuan Chu et al.
Brain research, 1343, 20-27 (2010-04-28)
Central dopaminergic system exerts profound modulation on spontaneous glutamate release in various brain regions mainly through D(1) receptor/cAMP/PKA pathway. It remains unclear whether the phosphatidylinositol (PI)-linked D(1)-like receptors are also involved in such modulatory actions. The identification of substituted phenylbenzazepine
Guang-Yu Li et al.
Molecular vision, 18, 2882-2895 (2012-12-13)
Oxidative stress is widely implicated in the death of retinal ganglion cells associated with various optic neuropathies. Agonists of the dopamine D(1) receptor have recently been found to be potentially neuroprotective against oxidative stress-induced injury. The goal of this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service