Skip to Content
Merck
All Photos(2)

Key Documents

S2071

Sigma-Aldrich

Anti-Sodium Channel NaV1.8 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-PN3, Anti-SCN10A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

immunohistochemistry (frozen sections): suitable using adult rat DRG
western blot: 1:200 using rat DRG lysates

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCN10A(6336)
mouse ... Scn10a(20264)
rat ... Scn10a(29571)

General description

Sodium voltage-gated channel α subunit 10 (SCN10A) codes for Nav1.8 sodium channels. It is expressed in nociceptive neurons. This gene is mapped to human chromosome 3p22.2.

Immunogen

peptide (C)EDEVAAKEGNSPGPQ corresponding to residues 1943-1957 of rat Nav1.8. This sequence has 14/15 residues identical in mouse and 7/15 residues identical in human.

Application

Anti-Sodium Channel NaV1.8 antibody has been used in immunohistochemistry and immunocytochemistry.

Biochem/physiol Actions

Sodium voltage-gated channel α subunit 10 (SCN10A) plays a major role in human pain. It is involved in the pathophysiology of cerebellar deficits in MS (multiple sclerosis).

Target description

Anti-Sodium Channel NaV1.8 specifically recognizes NaV1.8. Voltage-gated sodium channels (VGSCs) are present in most excitable cells. They play a crucial role in regulating the cell excitability, being primarily responsible for the

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, with 1% bovine serum albumin, and 0.05 % sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional NaV1. 8 Channels in Intracardiac NeuronsNovelty and Significance: The Link Between SCN10A and Cardiac Electrophysiology.
Verkerk AO, et al.
Circulation Research, 111(3), 333-343 (2012)
A SCN10A SNP biases human pain sensitivity.
Duan G, et al.
Molecular Pain, 12, 1744806916666083-1744806916666083 (2016)
Tomislav Kokotović et al.
Frontiers in molecular neuroscience, 14, 720973-720973 (2021-10-15)
PR domain-containing member 12 (PRDM12) is a key developmental transcription factor in sensory neuronal specification and survival. Patients with rare deleterious variants in PRDM12 are born with congenital insensitivity to pain (CIP) due to the complete absence of a subtype
Channelopathy-related SCN10A gene variants predict cerebellar dysfunction in multiple sclerosis.
Roostaei T, et al.
Neurology, 86(5), 410-417 (2016)
Common SCN10A variants modulate PR interval and heart rate response during atrial fibrillation.
Delaney JT, et al.
Europace, 16(4), 485-490 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service