Skip to Content
Merck
All Photos(1)

Key Documents

S2007

Sigma-Aldrich

Sera from goat

lyophilized powder (from clotted whole blood)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
biological source:
goat
origin:
USA origin
form:
lyophilized powder (from clotted whole blood)

biological source

goat

form

lyophilized powder (from clotted whole blood)

origin

USA origin

storage temp.

2-8°C

General description

Goat serum is applicable for a wide range of cell lines and primary cell cultures. It can be used as a supplement in growth media to culture primary cells obtained from guinea pig embryos, mouse peritoneal macrophages, monkey kidneys, chick embryos, and well-established cell lines. Serum provides essential nutritional and macromolecular growth factors for cell growth when added to the basal medium.

Application

Sera from goat has been used:

  • as a supplement to incubate mice ventricular cryosections for 4-hydroxynonenal (4HNEJ-2) immunostaining
  • to block unspecific antigens on reticulum cell sarcoma for immunofluorescence
  • as a component of the phosphate buffer saline (PBS) for incubation of slide-mounted sections for immunohistochemical analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicoletta Rizzi et al.
Neurobiology of disease, 114, 74-84 (2018-02-28)
The Parkinson's disease (PD) evolves over an extended period of time with the onset occurring long before clinical signs begin to manifest. Characterization of the molecular events underlying the PD onset is instrumental for the development of diagnostic markers and
Janus Rl Kobbersmed et al.
eLife, 9 (2020-02-23)
Chemical synaptic transmission relies on the Ca2+-induced fusion of transmitter-laden vesicles whose coupling distance to Ca2+ channels determines synaptic release probability and short-term plasticity, the facilitation or depression of repetitive responses. Here, using electron- and super-resolution microscopy at the Drosophila
Anton Baysa et al.
Basic research in cardiology, 110(2), 13-13 (2015-02-15)
Heart rupture and heart failure are deleterious complications of myocardial infarction. The ShcA gene encodes for three protein isoforms, p46-, p52- and p66ShcA. p66ShcA induces oxidative stress. We studied the role of p66ShcA post-infarction. Expression of p66ShcA was analyzed in
Smita Paranjape
Indian journal of experimental biology, 42(1), 26-35 (2004-07-28)
Serum is frequently added to the defined basal medium as a source of certain nutritional and macromolecular growth factors essential for cell growth. Although a number of synthetic media have been prepared serum continues to be used in cell culture
Majid N Humoud et al.
Journal of virology, 90(14), 6489-6501 (2016-05-06)
In response to stress such as virus infection, cells can stall translation by storing mRNAs away in cellular compartments called stress granules (SGs). This defense mechanism favors cell survival by limiting the use of energy and nutrients until the stress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service