Skip to Content
Merck
All Photos(1)

Key Documents

S1194

Sigma-Aldrich

SB-258585 dihydrochloride

≥98% (HPLC), powder

Synonym(s):

4-Iodo-N-[4-methoxy-3-(4-methylpiperazin-1-yl)phenyl]benzene-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22N3O3SI · 2HCl
Molecular Weight:
560.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

description

Store under nitrogen; package with desiccants at 4°C

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].COc1ccc(NS(=O)(=O)c2ccc(I)cc2)cc1N3CCN(C)CC3

InChI

1S/C18H22IN3O3S.2ClH/c1-21-9-11-22(12-10-21)17-13-15(5-8-18(17)25-2)20-26(23,24)16-6-3-14(19)4-7-16;;/h3-8,13,20H,9-12H2,1-2H3;2*1H

InChI key

QDYGVTFRTVLFCD-UHFFFAOYSA-N

Biochem/physiol Actions

5-HT6 serotonin receptor antagonist.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer C Roberts et al.
Brain research, 934(1), 49-57 (2002-04-09)
We used the highly selective 5-HT(6) receptor radioligand [(125)I]SB-258585 (4-iodo-N-[4-methoxy-3-(4-methylpiperazin-1-yl)phenyl]benzene-sulfonamide) to perform autoradiographic binding studies on the rat brain. High levels of specific binding occurred in the corpus striatum, nucleus accumbens, Islands of Calleja and the olfactory tubercle. A high
W D Hirst et al.
British journal of pharmacology, 130(7), 1597-1605 (2000-08-06)
SB-258585 (4-Iodo-N-[4-methoxy-3-(4-methyl-piperazin-1-yl)-phenyl]-benzen esulphonamide) is a high affinity ligand at 5-HT(6) receptors. It displays over 100 fold selectivity for the 5-HT(6) receptor over all other 5-HT receptors tested so far. SB-258585 has been radiolabelled, to high specific activity, for its characterization
C Routledge et al.
British journal of pharmacology, 130(7), 1606-1612 (2000-08-06)
SB-271046, potently displaced [(3)H]-LSD and [(125)I]-SB-258585 from human 5-HT(6) receptors recombinantly expressed in HeLa cells in vitro (pK(i) 8.92 and 9.09 respectively). SB-271046 also displaced [(125)I]-SB-258585 from human caudate putamen and rat and pig striatum membranes (pK(i) 8.81, 9.02 and
Wei Zhang et al.
Scientific reports, 8(1), 15753-15753 (2018-10-27)
Epidemiological observations have shown that schizophrenia patients after long-term drug treatment exhibited reduced tumor incidences. The potential anticancer effects of antipsychotic drugs are subsequently demonstrated. These drugs are of great interest as agents against untreatable brain metastases because of their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service