Skip to Content
Merck
All Photos(1)

Key Documents

S0417

Sigma-Aldrich

Silibinin

≥98% (HPLC)

Synonym(s):

2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-6-(3,5,7-trihydroxy-4-oxobenzopyran-2-yl)benzodioxin, Silybin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥98% (HPLC)

storage temp.

−20°C

SMILES string

COc1cc(ccc1O)[C@@H]2Oc3cc(ccc3O[C@H]2CO)[C@H]4Oc5cc(O)cc(O)c5C(=O)[C@H]4O

InChI

1S/C25H22O10/c1-32-17-6-11(2-4-14(17)28)24-20(10-26)33-16-5-3-12(7-18(16)34-24)25-23(31)22(30)21-15(29)8-13(27)9-19(21)35-25/h2-9,20,23-29,31H,10H2,1H3/t20-,23+,24-,25?/m0/s1

InChI key

SEBFKMXJBCUCAI-ILJKEPSESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silibinin, a principal component of silymarin, is a flavonolignan and an orally active flavonoid. It is isolated from the seeds of milk thistle (Sylibum marianum L).

Application

Silibinin has been used:
  • to study its effect on gene expression levels of various proteins involved in chromatin regulations of prostate cancer, by real time polymerase chain reaction (RT-PCR)
  • to study its effect on cell proliferation in platelet-derived growth factor (PDGF)-treated human tenon′s fibroblasts (HTFs) by investigating the expression of proliferating cell nuclear antigen (PCNA) and by water-soluble tetrazolium salt (WST-1) assay
  • to examine its effect on gene expression levels of stromelysine 1 (STM1), acetyl hexoseamines and collagen production during skin wound healing
  • to study its inhibitory effect on Escherichia coli ATP synthase

Biochem/physiol Actions

Silibinin functions as a hepatoprotectant in patients with acute and chronic liver injury. This flavonoid fights against various cancer cells including, prostate, skin, breast, colon, lung, bladder and hepatocellular carcinoma (HCC). Silibinin exhibits its efficacy as an anti-cancer agent by blocking the secretion of proangiogenic factors from tumor cells, and by hindering growth and inducing apoptotic death of endothelial cells. In addition, it also interrupts capillary tube formation on Matrigel. Silibinin might be a good candidate for chemoprevention of prostate cancer (PC). It might be used as a potential therapeutic regimen for the treatment of endometriosis in vitro and in vivo.

Components

Contains both A and B diastereomers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Inhibition of Kupffer cell functions as an explanation for the hepatoprotective properties of silibinin
Dehmlow C, et al.
Hepatology, 23(4), 749-754 (1996)
Silibinin-induced endoplasmic reticulum stress and mitochondrial dysfunction suppress growth of endometriotic lesions
Ham J, et al.
Journal of Cellular Physiology, 234(4), 4327-4341 (2019)
Prostate cancer prevention by silibinin
Singh RP, et al.
Current Cancer Drug Targets, 4(1), 1-11 (2004)
Zoe Mariño et al.
Journal of hepatology, 58(3), 415-420 (2012-10-16)
Hepatitis C recurrence after liver transplantation (LT) is the main problem of most transplant programs. We aimed at assessing the antiviral activity and safety of intravenous silibinin (SIL) administered daily during the peri-transplant period. This was a single-centre, prospective, randomized
A novel role of silibinin as a putative epigenetic modulator in human prostate carcinoma
Anestopoulos L, et al.
Molecules (Basel), 22(1), 62-62 (2017)

Articles

Chronic inflammation is an underlying factor in the development and progression of many of the chronic diseases of aging, such as arthritis, atherosclerosis, diabetes, and cancer.

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service