Skip to Content
Merck
All Photos(1)

Key Documents

RES1052B-B7

SAFC

BIOTIN

Synonym(s):

Biotin, D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Beilstein:
86838
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

97.5-100.5%

form

powder

impurities

Trace metals; tested

color

white

mp

231-233 °C (lit.)

suitability

suitable for manufacturing use

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to the documentation for this product requires a confidentiality disclosure agreement.

Packaging

Product is available in the following package sizes:
RES1052B-B701X: 1 g container
RES1052B-B707X: 10 g container
RES1052B-B708X: 25 g container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corey J Fugate et al.
Biochimica et biophysica acta, 1824(11), 1213-1222 (2012-02-14)
The enzyme cofactor and essential vitamin biotin is biosynthesized in bacteria, fungi, and plants through a pathway that culminates with the addition of a sulfur atom to generate the five-membered thiophane ring. The immediate precursor, dethiobiotin, has methylene and methyl
Pierre-Alexandre Driguez et al.
Natural product reports, 31(8), 980-989 (2014-04-08)
Covering: up to November 2013. Heparin and heparan sulfate are natural polysaccharides with strong structural variations, which are responsible for their numerous specific biological properties. One key target of heparin, among others, is antithrombin, a serine protease inhibitor that, upon
Joseph R Burgoyne et al.
Methods in enzymology, 473, 281-303 (2010-06-02)
The recent development of robust methods for the detection of proteins susceptible to S-nitrosylation (RSNO) and sulfenation (RSOH) has provided greater insight into the role of these oxidative modifications in cell signaling. These techniques, which have been termed "biotin-switch" methods
Shigehiro Takahashi et al.
Analytical and bioanalytical chemistry, 402(5), 1749-1758 (2011-08-26)
In this review, the preparation and properties of protein architectures constructed by layer-by-layer (LbL) deposition through avidin-biotin and concanavalin A (Con A)-sugar interactions are discussed in relation to their use for optical and electrochemical biosensors. LbL films can be constructed
Abdussalam Adina-Zada et al.
Biochemical Society transactions, 40(3), 567-572 (2012-05-24)
The activity of the biotin-dependent enzyme pyruvate carboxylase from many organisms is highly regulated by the allosteric activator acetyl-CoA. A number of X-ray crystallographic structures of the native pyruvate carboxylase tetramer are now available for the enzyme from Rhizobium etli

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service