Skip to Content
Merck
All Photos(1)

Key Documents

RAB1181

Sigma-Aldrich

Human LBP ELISA Kit

for cell culture supernatants, plasma, and serum samples

Synonym(s):

LBP, Lipopolysaccharide-binding protein 

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.77

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum sample(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.82 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LBP(3929)

General description

Human lipopolysaccharide-binding protein (LBP) ELISA (enzyme-linked immunosorbent assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of a target protein in biological samples, such as serum, plasma, cell culture supernatants, urine, and cell and tissue lysates. LBP is an acute-phase protein. It can initiate an immune response by binding to the bacterial cell wall. LBP is localized in the liver.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of lipopolysaccharide binding protein reveals the location of a frequent mutation that impairs innate immunity
Eckert J K, et al.
Immunity, 39(4), 647-660 (2013)
Tongai Gibson Maponga et al.
BMC infectious diseases, 18(1), 214-214 (2018-05-10)
Co-infection with HIV negatively impacts the progression of chronic hepatitis B virus (HBV) infection, including causing rapid progression to liver fibrosis. Sub-Saharan Africa represents arguably the most important intersection of high endemicity of both chronic hepatitis B virus (HBV) infection
Badreldin H Ali et al.
Biomolecules, 10(5) (2020-05-18)
We investigated some reproductive actions of hookah smoke (HS) exposure (30 min/day, for 30 days) in male mice, and the possible mitigative effect of the prebiotic agent gum acacia (GA) thereon. Control mice were air-exposed (AE). Twenty-four hours after the
Tao Liu et al.
mBio, 10(2) (2019-03-14)
Cyclophosphamide (CTX) is widely used in cancer chemotherapy, but it often induces mucositis, in which the disruption of the gut microbiota might play a pivotal role. Whether the manipulation of the gut microbiota can be used as a strategy to
Javier Lopez-Moreno et al.
Experimental gerontology, 109, 119-125 (2017-11-14)
Aging is an important determinant of the rate of atherosclerosis development, mainly through low-grade inflammation. Diet, and particularly its fat content, modulates the inflammatory response in fasting and postprandial states. We aimed to study the effects of dietary fat on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service