RAB1073
Human Osteocalcin ELISA Kit
for cell culture supernatants, plasma, and serum samples
Synonym(s):
Osteocalcin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum sample(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.4 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... BGLAP(632)
General description
Osteocalcin (OC/OCN) is a bone-building protein. This γ-carboxylated protein is made up of 49 amino acids and is synthesized by osteoblasts, odontoblasts and hypertrophic chondrocytes. The bone γ-carboxyglutamate protein (BGLAP), that produces OCN is located on human chromosome 1q22.
This ELISA antibody pair detects human Osteocalcin. Other species are not determined.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.
Please refer to the attached Protocolfor details.
Biochem/physiol Actions
Osteocalcin (OC/ OCN) plays a major role in the bone remodeling cycle, affecting bone mineralization. It is considered as a marker of bone formation. OCN guards against the progression of diabetes and the metabolic syndrome.
Other Notes
A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kevin D Cashman et al.
Nutrients, 11(8) (2019-07-31)
Reformulation of bread in terms of salt content remains an important measure to help achieve a reduction in salt intake in the population and for the prevention of hypertension and elevated blood pressure (BP). Our fundamental studies on the reduction
Jie Zhu et al.
Journal of diabetes and its complications, 31(8), 1354-1359 (2017-06-11)
This study presents nine patients with mitochondrial tRNA Leu (UUR) m.3243A>G mutation and compares the clinical characteristics and diabetes complications with type 1 diabetes (T1DM) or early onset type 2 diabetes (T2DM). The study covers 9 patients with MIDD, 33
Comprehensive metabolic characterization of serum osteocalcin action in a large non-diabetic sample
Entenmann L, et al.
Testing, 12(9), e0184721-e0184721 (2017)
Guangwen Jin et al.
FEBS open bio, 8(4), 523-532 (2018-04-11)
Osteoporosis is a progressive bone disease caused by an imbalance between bone resorption and formation. Recently, plasminogen activator inhibitor-1 (PAI-1) was shown to play an important role in bone metabolism using PAI-1-deficient mice. In this study, we evaluated the therapeutic
Analysis of osteocalcin as a candidate gene for type 2 diabetes (T2D) and intermediate traits in Caucasians and African Americans
Das S K, et al.
Disease Markers, 28(5), 281-286 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service