Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 28.7 pg/mL
standard curve range: 28.67-7000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... LGMN(5641)
General description
The antibody pair provided in this kit recognizes human Legumain.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cancer biology & therapy, 18(4), 222-228 (2017-03-10)
Epithelial ovarian cancer (EOC) is the most common and lethal cancer-related death among females in the world. Asparaginyl endopeptidase (AEP) is a member of C13 family peptidases and expressed in the extracellular matrix and tumor cells. The aim of this
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(1), 193-210 (2020-11-12)
Oral squamous cell carcinoma (OSCC) is one of the most painful cancers, which interferes with orofacial function including talking and eating. We report that legumain (Lgmn) cleaves protease-activated receptor-2 (PAR2) in the acidic OSCC microenvironment to cause pain. Lgmn is
Basic & clinical pharmacology & toxicology, 125(2), 89-99 (2019-03-28)
Proton pump inhibitors (PPIs) are prodrugs used in the treatment of peptic ulcer diseases. Once activated by acidic pH, the PPIs subsequently inhibit the secretion of gastric acid by covalently forming disulphide bonds with the SH groups of the parietal
Cellular oncology (Dordrecht), 43(2), 263-277 (2020-02-23)
Epithelial ovarian cancer (EOC) is one of the most malignant cancers in the gynecologic system. Many patients are diagnosed at an advanced stage with disseminated intra-peritoneal metastases. EOC spreads via both direct extension and trans-coelomic spread. However, the interplay between
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service