Skip to Content
Merck
All Photos(1)

Key Documents

RAB0695

Sigma-Aldrich

Human IL21 / Interleukin-21 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 110 pg/mL
standard curve range: 125-8000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL21(59067)

General description

Interleukin-21 (IL-21) is a pleiotropic cytokine. It belongs to the common γ-chain cytokines with broad pleiotropic actions. This gene is located on human chromosome 4q27.

Application

Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Interleukin-21 (IL-21) regulates chronic viral infection, thereby plays a major role for immune responses in a mouse model of HBV (hepatitis B virus) infection. This gene is linked to celiac disease, rheumatoid arthritis (RA), type 1 diabetes mellitus (type 1 DM) and psoriatic arthritis. IL-21 has the ability to promote maturation and increase the cytotoxicity of CD8+ T cells and natural killer (NK) cells.
The antibody pair provided in this kit recognizes Human Interleukin-21.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yolanda P Alemán-García et al.
Journal of immunology research, 2021, 6630715-6630715 (2021-03-26)
Systemic lupus erythematosus is characterized by high levels of IgG class autoantibodies that contribute to the pathophysiology of the disease. The formation of these autoantibodies occurs in the germinal centers, where there is cooperation between follicular T helper cells (TFH)
Identification of the novel 3? UTR sequences of human IL-21 mRNA as potential targets of miRNAs
Enomoto Y, et al.
Scientific Reports, 7(1), 7780-7780 (2017)
Association of the autoimmunity locus 4q27 with juvenile idiopathic arthritis
Albers HM, et al.
Arthritis and Rheumatism, 60(3), 60(3)-60(3) (2009)
Induction of IL-21 in Peripheral T Follicular Helper cells is Indicator of Influenza Vaccine Response in Previously Vaccinated HIV-Infected Pediatric Cohort
de Armas LR, et al.
Journal of Immunology (2017)
Biological effects of IL-21 on different immune cells and its role in autoimmune diseases.
Gharibi T, et al.
Immunobiology, 221(2), 357-367 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service