Skip to Content
Merck
All Photos(1)

Documents

RAB0568

Sigma-Aldrich

Bovine INS / Insulin ELISA Kit

Synonym(s):

Insulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

bovine

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4.5 μIU/mL
standard curve range: 4.69-300 μIU/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

bovine ... INS(280829)

General description

The antibody pair provided in this kit recognizes Bovine Insulin.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Guang Li et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 97, 1229-1237 (2017-11-18)
Obstructive sleep apnea hypopnea syndrome (OSAHS) is associated with glucose intolerance, insulin resistance and type 2 diabetes mellitus (T2DM). Although several studies have revealed that intermittent hypoxia (IH) in OSAHS may further aggravate pancreatic β cell damage and promote the
Oswaldo Cifuentes-Lopez et al.
Life (Basel, Switzerland), 11(6) (2021-07-03)
The objective of this experiment was to evaluate the effects of feeding different levels concentrations of dietary calcium propionate (CaPr) on lambs' growth performance; ruminal fermentation parameters; glucose-insulin concentration; and hypothalamic mRNA expression for neuropeptide Y (NPY), agouti-related peptide (AgRP)
Manuel Blandino-Rosano et al.
Nature communications, 8, 16014-16014 (2017-07-13)
Deregulation of mTOR complex 1 (mTORC1) signalling increases the risk for metabolic diseases, including type 2 diabetes. Here we show that β-cell-specific loss of mTORC1 causes diabetes and β-cell failure due to defects in proliferation, autophagy, apoptosis and insulin secretion
Xiaoying Xing et al.
Drug delivery, 25(1), 23-34 (2017-12-12)
Surfactin, a lipopeptide produced by Bacillus species, has been used for the oral delivery of insulin. In this study, another lipopeptide of iturin was tested for its ability to orally delivery insulin alone or plus surfactin. Iturin could form co-precipitate
Xiangyu Gao et al.
International heart journal, 59(2), 272-278 (2018-02-16)
Circulating relaxin (RLX) is altered in patients with diabetes mellitus (DM) or cardiovascular diseases. This study was designed to evaluate the changes of RLX in patients with unstable angina (UA) complicated with various categories of abnormal glucose metabolism.Patients who confirmed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service