Skip to Content
Merck
All Photos(2)

Key Documents

RAB0462

Sigma-Aldrich

Human β IG-H3 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

Quality Level

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 25 pg/mL
standard curve range: 24.58-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TGFBI(7045)

General description

The Human β IG-H3 (also known as Kerato-epithelin, RGD- CAP) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human β IG-H3 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human β-IGH3

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Zhu et al.
International journal of clinical and experimental pathology, 8(1), 702-710 (2015-03-11)
Transforming growth factor-beta-induced (TGFBI) serves as a linker protein and plays a role in the activation of morphogenesis, cell proliferation, adhesion, migration, differentiation and inflammation. High expression levels of the human TGFBI gene are correlated with numerous human malignancies. In
Laura Lauden et al.
The Journal of investigative dermatology, 134(6), 1675-1685 (2014-02-07)
Tumor-produced extracellular matrix (ECM) proteins can be key elements in tumor growth and metastasis. Transforming growth factor beta-inducible (TGFBI) protein is a secreted ECM component that can have dual function in cancer, acting as tumor suppressor or promoter. Although TGFBI
Shih Sheng Jiang et al.
Biochimica et biophysica acta, 1842(7), 1080-1087 (2014-03-26)
The nonsense mutations of the hepatitis B virus (HBV) surface (S) gene have been reported to have oncogenic potential. We have previously identified several transforming nonsense mutations of the HBV S gene from hepatocarcinoma (HCC) patients. Among them, the sW182*

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service