Skip to Content
Merck
All Photos(2)

Key Documents

RAB0408

Sigma-Aldrich

Mouse Prolactin ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 30 pg/mL
standard curve range: 27.43-20000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Prl(19109)

General description

The Mouse Prolactin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse Prolactin in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse Prolactin

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Prolactin (Prl) is processed to lower molecular weight N-terminal fragments, 14-18kDa, called vasoinhibins, which have antiangiogenic effects. In endothelial cells, the 16kDa prolactin form exerts antiproliferative and proapoptotic effects which result in cell cycle arrest, caspase and NFκB (nuclear factor) activation and regulation of Bcl-2 protein family. In sexually mature female mice, the plasma levels of prolactin remain constant throughout estrous cycle, but show a sudden increase during proestrous cycle due to increase in circulating levels of estrogens. Studies show that Prl activates T cells, B cells, natural killer (NK) cells, macrophages, neutrophils and dendritic cells and there is a significant level of hyperprolactinemia incidence in patients with autoimmune diseases, such as rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). Studies in rats show that Prl is involved in T cell homing to mammary glands during early lactation and that hypoprolactemia during lactation results in modified T cell distribution with increased T cell in circulation and reduced in mammary glands.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impaired mammary gland T cell population during early lactation in hypoprolactinemic lactation-deficient rats.
Mackern-Oberti JP
Reproduction (Cambridge, England) null
N-terminal prolactin-derived fragments, vasoinhibins, are proapoptoptic and antiproliferative in the anterior pituitary.
Ferraris J
PLoS ONE null
Conditional Deletion of the Prolactin Receptor to Identify Brain-Specific Actions of Prolactin
3 Biotech null
Daniella C Buonfiglio et al.
Hormones and behavior, 71, 60-68 (2015-04-22)
Prolactin and placental lactogens control mammary development and lactation as well as play an important role in maternal behaviors. However, the molecular mechanisms in the brain responsible for this regulation remain largely unknown. Therefore, the present study investigated whether Signal
A N Johnston et al.
Breast cancer research : BCR, 20(1), 42-42 (2018-05-21)
Psychiatric medications are widely prescribed in the USA. Many antipsychotics cause serum hyperprolactinemia as an adverse side effect; prolactin-Janus kinase 2 (JAK2)-signal transducer and activator of transcription 5 (STAT5) signaling both induces cell differentiation and suppresses apoptosis. It is controversial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service