Skip to Content
Merck
All Photos(1)

Key Documents

RAB0377

Sigma-Aldrich

Visfatin EIA Kit

for serum, plasma, culture supernatant and cell lysates

Synonym(s):

Visfatin ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

species reactivity

rat, human, mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

input

sample type serum
sample type culture supernatant(s)
sample type cell lysate
sample type plasma

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 0.778 ng/mL
standard curve range: 0.1-1000 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NAMPT(10135)

General description

The Visfatin Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting Visfatin peptide based on the principle of Competitive Enzyme Immunoassay.

Visfatin, also known as nicotinamide phosphoribosyltransferase (Nampt)/ pre-B Cell colony enhancing factor is encoded by the gene mapped to human chromosome 7q22.3. The gene contains 11 exons encompassing 34.7kb. The encoded protein is an adipocytokine secreted by visceral fat.

Immunogen

Visfatin, synthetic peptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Visfatin EIA Kit has been used to measure the concentration of visfatin in blood samples.

Biochem/physiol Actions

Visfatin has a role in mammalian nicotinamide adenine dinucleotide (NAD+) biosynthesis. Studies have shown that mice deficient in visfatin in the forebrain excitatory neurons show hyperactivity, diminished anxiety-like behaviors and impaired learning and memory. It has also been shown that visfatin stimulates glucose uptake and adenosine monophosphate-activated protein kinase (AMPK) phosphorylation in mouse C2C12 skeletal muscle cells. Visfatin regulates inflammation and insulin sensitivity.

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A visfatin promoter polymorphism is associated with low?grade inflammation and type 2 diabetes.
Zhang Y-Y, et al.
Obesity (Silver Spring, Md.), 14(12), 2119-2126 (2006)
Matthaios G Pitoulias et al.
Cytokine, 91, 140-144 (2017-01-11)
Visfatin is an adipokine molecule acting as an essential coenzyme in multiple cellular redox reactions. The increased serum levels of Visfatin have been correlated with metabolic syndrome and endothelial homeostasis. In this study we investigate the possible relationship of Visfatin
Effects of Diet and Exercise-Induced Weight Loss in Sedentary Obese Women on Inflammatory Markers, Resistin, and Visfatin.
Khanna D, et al.
Obesity and Nutrition of J, 102 (2017)
Single and Concurrent Effects of Endurance and Resistance Training on Plasma Visfatin, Insulin, Glucose and Insulin Resistance of Non-Athlete Men with Obesity.
Tayebi S M, et al.
Annals of applied sport science., 4(4), 21-31 (2016)
Ashleigh M Philp et al.
International journal of molecular sciences, 22(13) (2021-07-03)
Obesity increases the risk of hip osteoarthritis (OA). Recent studies have shown that adipokine extracellular nicotinamide phosphoribosyltransferase (eNAMPT or visfatin) induces the production of IL-6 and matrix metalloproteases (MMPs) in chondrocytes, suggesting it may promote articular cartilage degradation. However, neither

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service