Skip to Content
Merck
All Photos(2)

Key Documents

RAB0293

Sigma-Aldrich

Human IFNL1 / Interferon lambda-1 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

Cytokine Zcyto21, IFN-lambda-1, IL-29, IL29, Interleukin-29, ZCYTO21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 13.72-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL29(282618)

General description

The Human IL-29 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-29 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IL-29

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bogusz Falkowski et al.
Mediators of inflammation, 2020, 4748612-4748612 (2020-07-23)
The most frequent cause of hyperthyroidism is Graves' disease (GD). Orbitopathy is the most prevalent and recognizable extrathyroidal manifestation of Graves' disease with unrevealed pathogenesis. Interleukin 29 (IL-29) is a relatively newly discovered inflammatory cytokine. Thus, the aim of this
Tingshuang Xu et al.
Clinical rheumatology, 39(12), 3797-3804 (2020-05-30)
We aimed to investigate whether Interleukin-29 (IL-29) directly affects T follicular helper (Tfh) cell frequency in rheumatoid arthritis (RA), which are both related to RA-specific antibody responses. Here, we explored the effect of IL-29 on Tfh cell production in RA
Bogusz Falkowski et al.
Journal of clinical medicine, 9(1) (2020-01-16)
Type 1 diabetes mellitus (T1DM) is associated with chronic complications, which are the result of neurovascular changes. There is still a lack of universal biochemical markers of microvascular damage. The present study aimed to investigate whether selected inflammatory proteins are
Michael Sörensen et al.
Frontiers in immunology, 11, 96-96 (2020-03-03)
Cystic fibrosis (CF) patients frequently acquire Pseudomonas aeruginosa infections that have been associated with a bad prognosis and an increased rate of pulmonary exacerbations. Respiratory viruses can cause exacerbations in chronic pulmonary diseases including COPD or asthma and have been
Elisa Vintiñi et al.
Iranian journal of immunology : IJI, 14(4), 325-339 (2017-12-26)
Polyinosinic:polycytidylic acid (Poly-IC) has been used as a viral stimulus to mimic in vivo and in vitro infection induced by some viruses. To determine whether non-viable Lactobacillus casei CRL431 (LcM) can modulate the immune response induced by Poly I:C in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service