Skip to Content
Merck
All Photos(2)

Key Documents

RAB0037

Sigma-Aldrich

Human Procalcitonin ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

description

for cell culture supernatant and urine. Note :The detection range of this kit is 30 pg/ml - 20000 pg/ml.  Procalcitonin is low in normal serum and may be undetectable with this assay.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 30 pg/mL
standard curve range: 27.43-20000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CALCA(796)

General description

The Human Procalcitonin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Procalcitonin in cell culture supernatants and urine. Note: The detection range of this kit is 30 pg/ml - 20000 pg/ml.  Procalcitonin is low in normal serum and may be undetectable with this assay.

Immunogen

Recombinant Human Procalcitonin

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Procalcitonin or calcitonin related polypeptide a is a 116-amino acid precursor of calcitonin. It is secreted by monocytes activated by endotoxins or pathogens. The protein is a biomarker for sepsis and bacterial infections. Procalcitonin has been associated with hepatocyte dysfunction.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADDELISA 5X Assay/Sample Diluent Buffer D (Item K)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Procalcitonin--a Marker for Anastomotic Insufficiency after Pancreatoduodenectomy?
Lenschow C
Clinical Laboratory (2016)
Amedeo Amedei et al.
Internal and emergency medicine, 9(3), 303-309 (2012-10-12)
Helicobacter pylori infection is characterized by an inflammatory infiltrate, consisting mainly of neutrophils and T cells. This study was undertaken to evaluate the type of gastric T cell response elicited by the secreted peptidyl prolyl cis, trans-isomerase of H. pylori
F R Tehrani et al.
Climacteric : the journal of the International Menopause Society, 17(5), 591-597 (2014-04-11)
Despite the wide use of anti-Müllerian hormone (AMH) measurement as a clinical marker for assessment of ovarian reserve, a population-based estimate for its reference values is not available. In this study, we have estimated age-specific AMH levels in a large
Weiwei Zhu et al.
Biology of reproduction, 91(1), 8-8 (2014-05-31)
The mammalian testis is an immunoprivileged organ where local tissue-specific cells acquire an effective innate immune function against invading microbial pathogens. The present study demonstrated that mouse Leydig cells had innate antiviral activities in response to viral DNA challenge through
Procalcitonin Impairs Liver Cell Viability and Function In Vitro: A Potential New Mechanism of Liver Dysfunction and Failure during Sepsis?
Martin Sauer
BioMed Research International (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service