Skip to Content
Merck
All Photos(1)

Key Documents

R9259

Texan reagent reservoir for multichannel pipettes

without lid, non-sterile, pack of 100 ea

Synonym(s):

multichannel pipette reservoir, reagent reservoir

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121522
NACRES:
NB.01

material

V-bottom bottom
colorless
polypropylene

sterility

non-sterile

packaging

pack of 100 ea

manufacturer/tradename

Excel Scientific RSV-0175-N10

capacity

175 mL

application(s)

clinical testing

Looking for similar products? Visit Product Comparison Guide

General description

Texan reagent reservoirs are reusable -- an environmentally sound alternative to disposable reagent reservoirs. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste. Lids are removable -- attached with ball and socket hinges -- and latch tightly for secure storage of left-over reagents. With lids closed, Texan reagent reservoirs are stackable. A perimeter ridge on the lid prevents sliding. Construction is clear, chemical-resistant, autoclavable polypropylene. Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 mL.

Legal Information

Texan is a trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service