Skip to Content
Merck
All Photos(1)

Key Documents

R5404

Sigma-Aldrich

Monoclonal Anti-Rab9 antibody produced in mouse

~2 mg/mL, clone Rab9-29, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Rab9-29, monoclonal

form

buffered aqueous solution

mol wt

~23 kDa

species reactivity

human, rat, hamster, canine, mouse

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 2 μg/mL using total cell extract of HeLa cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB9A(9367)
mouse ... Rab9(56382)
rat ... Rab9a(84589)

General description

Monoclonal Anti-Rab9 (mouse IgG1 isotype) is derived from the hybridoma Rab9-29 produced by the fusion of mouse cells (NS1 cells) and splenocytes from BALB/c mice. Ras-related protein Rab-9A (Rab9) is a member of the Rab family of small guanosine triphosphatases (GTPases). The gene Rab9 is mapped to human chromosome Xp22.2. Rab9 GTPase is localized predominantly to late endosomes.

Specificity

Monoclonal Anti-Rab9 recognizes human, canine, rat, mouse and hamster Rab9 (approx. 23 kDa)

Immunogen

synthetic peptide corresponding to amino acids 187-200 of human Rab9, conjugated to KLH.

Application

Monoclonal Anti-Rab9 antibody produced in mouse has been used in:
  • western blotting
  • enzyme-linked immunosorbent assay(ELISA)
  • immunoprecipitation
  • immunocytochemistry

Biochem/physiol Actions

Ras-related protein Rab-9A (Rab9) guanosine triphosphatase (GTPase) is required for the transport of mannose 6-phosphate receptors (MRPs) from endosomes to the trans-Golgi network (TGN). Rab GTPases are central regulators of membrane trafficking between the different subcellular compartments of the eukaryotic cell. Rab9 GTPase in its GTP bound conformation binds specifically to an effector protein named p40 that stimulates the transport of mannose 6-phosphate receptors (MPRs) from endosomes to the TGN. Rab9 also interacts with the vesicle cargo selection protein tail-interacting protein of 47 kDa (TIP47). This interaction is required for Rab9 stability on late endosomes and may be crucial for vesicular transport in the virus life cycle.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel P Kloer et al.
The Journal of biological chemistry, 285(10), 7794-7804 (2010-01-06)
The Hermansky-Pudlak syndrome (HPS) is a genetic hypopigmentation and bleeding disorder caused by defective biogenesis of lysosome-related organelles (LROs) such as melanosomes and platelet dense bodies. HPS arises from mutations in any of 8 genes in humans and 16 genes
Laurie K Jackson et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(37), 14141-14146 (2008-09-13)
Salmonella's success at proliferating intracellularly and causing disease depends on the translocation of a major virulence protein, SifA, into the host cell. SifA recruits membranes enriched in lysosome associated membrane protein 1 (LAMP1) and is needed for growth of Salmonella
Ana Kucera et al.
Communicative & integrative biology, 9(4), e1204498-e1204498 (2016-08-31)
The small GTPase Rab9 has long been described as a protein that mediates endosome-to-trans-Golgi Network (TGN) transport, and specifically mannose-6-phospate receptor (MPR) recycling. However, studies have challenged this view by showing that Rab9 also is connected to sorting pathways toward
Daniela Ploen et al.
European journal of cell biology, 92(12), 374-382 (2014-02-01)
Hepatitis C virus (HCV) morphogenesis and release are closely linked to lipid metabolism. It has been described recently by our group that TIP47 plays an essential role for the targeting of the NS5A-complexed RNA genome from the replicon complex to
X Ke et al.
Annals of human genetics, 67(Pt 3), 242-249 (2003-08-14)
Sequence comparison analyses have been carried out for 19 genes escaping X-inactivation versus 73 genes subject to X-inactivation, and 100 randomly chosen X chromosome genes versus 100 randomly chosen autosomal genes. The coding sequence of the genes and their upstream

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service