R3380
Ribonucleoside vanadyl complexes
200 mM
Synonym(s):
vanadyl ribonucleoside complexes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
Ribonucleoside vanadyl complexes are stable complexes formed between ribonucleotides. They are low molecular weight inhibitors of cellular ribonucleases (RNases) that are used for the isolation of RNA from cells.
Application
Ribonucleoside vanadyl complexes are used as ribonuclease inhibitors during cell lysis and mRNA purification, to protect mRNA during cDNA production, and to protect RNA during digestion of DNA by DNaseI.
Ribonucleoside vanadyl complexes has been used:
- as a component of lysis buffer for cell lines prior to Poly(U) pull down assay
- to inhibit nucleases prior to immunoprecipitation (IP) of m6A-containing RNA fragment
- as a component of hybridization buffer for human retinal slices
Biochem/physiol Actions
Ribonucleoside vanadyl complexes (RVC) elicits antimicrobial functionality by inhibiting ribosomal subunit formation and suppressing the growth of Staphylococcus aureus and Escherichia coli. It is also used as a preservative solution for tissues.
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Inhibition of intractable nucleases with ribonucleoside--vanadyl complexes: isolation of messenger ribonucleic acid from resting lymphocytes.
Biochemistry, 18(23), 5143-5149 (1979-11-13)
Effect of ribonucleoside-vanadyl complexes on enzyme-catalyzed reactions central to recombinant deoxyribonucleic acid technology.
Biochemistry, 21(19), 4602-4608 (1982-09-14)
Enzymology Primer for Recombinant DNA Technology (1996)
Neuroinflammatory processes, A1 astrocyte activation, and protein aggregation in the retina of Alzheimer's Disease patients, possible biomarkers for early diagnosis.
Frontiers in Neuroscience, 13, 925-925 (2019)
Transcriptome-wide mapping of N6-methyladenosine by m6A-Seq
Test, 560, 131-147 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service