Skip to Content
Merck
All Photos(1)

Key Documents

PZ0002

Sigma-Aldrich

Maraviroc

≥98% (HPLC)

Synonym(s):

4,4-Difluoro-N-[(1S)-3-[(3-exo)-3-[3-methyl-5-(1-methylethyl)-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]oct-8-yl]-1-phenylpropyl]cyclohexanecarboxamide, UK-427857

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H41F2N5O
CAS Number:
Molecular Weight:
513.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]-15/D

color

white

solubility

DMSO: >30 mg/mL

storage temp.

room temp

SMILES string

CC(C)c1nnc(C)n1[C@H]2C[C@@H]3CC[C@H](C2)N3CC[C@H](NC(=O)C4CCC(F)(F)CC4)c5ccccc5

InChI

1S/C29H41F2N5O/c1-19(2)27-34-33-20(3)36(27)25-17-23-9-10-24(18-25)35(23)16-13-26(21-7-5-4-6-8-21)32-28(37)22-11-14-29(30,31)15-12-22/h4-8,19,22-26H,9-18H2,1-3H3,(H,32,37)/t23-,24+,25-,26-/m0/s1

InChI key

GSNHKUDZZFZSJB-QYOOZWMWSA-N

Gene Information

human ... CCR5(1234)

Application

Maraviroc has been used as a C-C chemokine receptor type 5 (CCR5-specific) inhibitor in murine monocyte/macrophage cell line J774A.1, porcine uterine luminal epithelial cells and human lung CD4+ T cells.

Biochem/physiol Actions

Maraviroc is a CCR5 chemokine receptor antagonist and antiretroviral entry inhibitor.
Maraviroc is a CCR5 chemokine receptor antagonist, and antiretroviral entry inhibitor. Maraviroc inhibits HIV entry by blocking interaction of viral coat protein gp120 with the receptor.
Maraviroc is metabolised by cytochrome P450 3A4. The transport and absorption of maraviroc in the intestine and bile is regulated by P-glycoprotein (Pgp). Maraviroc binds to the active site pocket of chemokine receptor CCR5 making the receptor inactive.

Features and Benefits

This compound is featured on the Chemokine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan G Sierra-Madero et al.
The lancet. HIV, 1(2), e60-e67 (2015-10-02)
Immune reconstitution inflammatory syndrome (IRIS) is a common complication of antiretroviral therapy (ART) in patients with HIV. IRIS is associated with an increased risk of admission to hospital and death. We assessed whether CCR5 blockade with maraviroc reduces the risk
Assessment of the absorption, metabolism and absolute bioavailability of maraviroc in healthy male subjects
Abel S, et al.
British Journal of Clinical Pharmacology, 65, 60-67 (2008)
Laura Pérez-Martínez et al.
The Journal of antimicrobial chemotherapy, 69(7), 1903-1910 (2014-03-22)
Non-alcoholic fatty liver disease (NAFLD) is the most common cause of chronic liver disease in the general population. The NAFLD spectrum ranges from simple steatosis to cirrhosis. The chemokine CCL5/RANTES plays an important role in the progression of hepatic inflammation
Procoagulant and proinflammatory effects of red blood cells on lipopolysaccharide-stimulated monocytes
Osterud B, et al.
Journal of Thrombosis and Haemostasis, 13(9), 1676-1682 (2015)
Structure of the CCR5 chemokine receptor-HIV entry inhibitor maraviroc complex
Tan Q, et al.
Science, 341(6152), 1387-1390 (2013)

Articles

We offer many products related to chemokine receptors for your research needs.

We offer many products related to chemokine receptors for your research needs.

We offer many products related to chemokine receptors for your research needs.

We offer many products related to chemokine receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service