Skip to Content
Merck
All Photos(3)

Key Documents

PRS2501

Sigma-Aldrich

Anti-CIKS antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-TRAF3 Interacting protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 63 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

CIKS (connection to IKK (IκB kinase) and SAPK (stress-activated protein kinase) /JNK (jun kinase)) is a cytoplasmic protein, that exists in the nucleus. It is also termed as Act1(NF-κB activator 1) and TRAF3IP2 (TRAF3 interacting protein 2). It is located on chromosome 6q21.

Immunogen

a synthetic peptide corresponding to amino acids 2-15 of human CIKS.

Biochem/physiol Actions

CIKS (connection to IKK (IκB kinase) and SAPK (stress-activated protein kinase) /JNK (jun kinase)) is susceptible to various diseases like psoriasis and psoriatic arthritis. It is responsible for the cardiomyocyte growth, fibroblast migration and proliferation, promoted by aldosterone. It participates in the signaling function of IL-17 (interleukin) cytokine family. It can act as a transcriptional enhancer due to its presence in the nuclear compartment of airway epithelial cells.

Linkage

The action of this antibody can be blocked using blocking peptide SBP2501.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Novel Nuclear Function for the Interleukin-17 Signaling Adaptor Protein Act1
Velichko S, et al.
PLoS ONE, 11(10) (2016)
Identification of alternatively spliced Act1 and implications for its roles in oncogenesis
Xia YF, et al.
Biochemical and Biophysical Research Communications, 296(2), 406-412 (2002)
Aldosterone-induced cardiomyocyte growth, and fibroblast migration and proliferation are mediated by TRAF3IP2
Somanna NK, et al.
Cellular Signalling, 27(10), 1928-1938 (2015)
A pharmacogenetics study in Mozambican patients treated with nevirapine: full resequencing of TRAF3IP2 gene shows a novel association with SJS/TEN susceptibility
Ciccacci C, et al.
International Journal of Molecular Sciences, 16(3), 5830-5838 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service