Skip to Content
Merck
All Photos(1)

Key Documents

PROTRA

Millipore

ProteoPrep® Reduction and Alkylation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

shelf life

≥1 yr (when stored at 2-8°C)

Quality Level

storage temp.

2-8°C

Application

The ProteoPrep Reduction & Alkylation Kit contains the necessary reagents to conveniently reduce and alkylate disulfide binds in preparation for 2D gel electrophoresis. These reagents are compatible with chaotropic extraction reagents and are conveneintly packaged to save time and increase efficiency. The kit contains five vials of both the reducing agent, tributylphosphine (TBP), and the alkylating agent, iodoacetamide.

Features and Benefits

  • Tributylphosphine is supplied safely as a 200 mM solution in N-methyl-2-pyrrolidine
  • Reduction and alkylation of protein samples increase 2D spot resolution
  • Conveniently packaged components save time and increase efficiency
  • Compatibility with chaotropic extraction reagents simplifies sample preparation

Legal Information

ProteoPrep is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shikha Shikha et al.
Microorganisms, 10(5) (2022-05-29)
Mitochondrial ribosomes are fundamental to mitochondrial function, and thus survival, of nearly all eukaryotes. Despite their common ancestry, mitoribosomes have evolved divergent features in different eukaryotic lineages. In apicomplexans, the mitochondrial rRNA is extremely fragmented raising questions about its evolution
Shinobu Takagi et al.
Bioscience, biotechnology, and biochemistry, 84(11), 2241-2252 (2020-07-23)
A simple intracellular proteomic study was conducted to investigate the biological activities of Aspergillus niger during industrial enzyme production. A strain actively secreting a heterologous enzyme was compared to a reference strain. In total, 1824 spots on 2-D gels were

Articles

This discussion will highlight some of these methodologies, namely, the use of Multiple Reaction Monitoring (MRM) and Protein-AQUA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service