Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.56
UNSPSC Code:
41106500
Storage temp.:
2-8°C
Shipped in:
ambient
Application
ProteoPrep® Blue Albumin & IgG Depletion Kit has been used:
- to deplete serum samples from immunoglobulin (Ig) G and albumin
- for albumin removal
- for albumin and IgG removal
- to deplete abundant plasma proteins for two-dimensional electrophoresis
Biochem/physiol Actions
ProteoPrep blue albumin depletion kit is designed to specifically remove albumin and IgG from 25 samples of human serum (25μL to 100μL) in preparation for two-dimensional electrophoresis (2DE). Albumin-depleted serum samples generated by this kit are in urea rather than salt-based buffers with little dilution, eliminating the need to precipitate the sample prior to performing 2DE.
Legal Information
ProteoPrep is a registered trademark of Merck KGaA, Darmstadt, Germany
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Anna Tomascova et al.
General physiology and biophysics, 38(4), 305-314 (2019-06-21)
Blood biomarkers are usually present in low concentration and can be masked by the high-abundance proteins, of which albumin is the predominant one. The purpose of this study was to compare four different albumin removal methods compatible with in-gel based
Zeyu Sun et al.
Theranostics, 9(4), 1200-1214 (2019-03-15)
Chronic HBV infection (CHB) can lead to acute-on-chronic liver failure (HBV-ACLF) characterized by high mortality. This study aimed to reveal ACLF-related proteomic alterations, from which protein based diagnostic and prognostic scores for HBV-ACLF were developed. Methods: Ten healthy controls, 16
Yiting Li et al.
Methods in molecular biology (Clifton, N.J.), 818, 81-96 (2011-11-16)
The development of a sensitive, specific, and non-invasive approach for cancer detection will facilitate early detection and, hence, improve the outcome of individuals with known cancer predispositions. Proteomic profiling of blood emerges to be a logical choice of such non-invasive
Global Trade Item Number
| SKU | GTIN |
|---|---|
| PROTBA-1KT | 04061836686963 |




