Skip to Content
Merck
All Photos(3)

Documents

PF0100

Sigma-Aldrich

Protease Fluorescent Detection Kit

high sensitivity assay

Synonym(s):

Protease Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

shipped in

dry ice

storage temp.

−20°C

Application

Protease Fluorescent Detection Kit has been used:
  • in the detection of proteolytic activity using casein based protease assay
  • for testing the extracellular proteolytic activity of fungal strains
  • for quantifying mitochondrial protease activity
  • for measuring the protease production from soil solution

Suitability

Detection limit: approx. 50 ng trypsin

Quantity

Supplied with sufficient reagents for up to 200 one mL assays.
Supplied with sufficient reagents for up to 200 one mL assays.

The trichloroacetic acid (TCA) component is provided as 6.1 N TCA solution. 0.6 N TCA Solutions may be prepared by a 10-fold dilution of the desired volume of T0699 with ultrapure water. It is not necessary to dilute the entire volume of T0699 for immediate use of the kit.

Analysis Note

Tested for detection of all four proteolytic classes (serine, apartic, cysteine and metalloproteinases).

Kit Components Only

Product No.
Description

  • Incubation buffer 25 mL

  • FITC-casein solution 5 mL

  • Assay buffer 200 mL

  • Protease standard (trypsin) 20 μg

  • FITC control 50 mg

  • Trichloroacetic acid (TCA) solution 30 mL

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Martin H Kunzmann et al.
Molecular bioSystems, 8(11), 3061-3067 (2012-09-20)
α-Alkylidene-γ-butyrolactones are quite common in nature and exhibit a broad spectrum of biological activities. We therefore synthesized a small library of xanthatine inspired α-alkylidene-γ-butyrolactones to screen non-pathogenic and uropathogenic E. coli strains by activity based protein profiling (ABPP). The identified
Contrasting effects of increased carbon input on boreal SOM decomposition with and without presence of living root system of Pinus sylvestris L.
Linden, A., et al.
Plant and Soil, 377, 145-158 (2014)
Joseph S Rom et al.
Virulence, 8(8), 1776-1790 (2017-09-15)
Staphylococcus aureus causes acute and chronic forms of infection, the latter often associated with formation of a biofilm. It has previously been demonstrated that mutation of atl, codY, rot, sarA, and sigB limits biofilm formation in the USA300 strain LAC
Caveolin-1 controls mitochondrial function through regulation of m-AAA mitochondrial protease
Volonte D, et al.
Aging (Albany. NY.), 8(10), 2355-2355 (2016)
Qiang Fu et al.
Radiation research, 175(6), 728-735 (2011-05-03)
The somatostatin analog SOM230 has potent radioprophylactic and radiation mitigating properties that are unrelated to cytoprotection but appear to be due to suppression of secretion of pancreatic enzymes into the intestinal lumen. To determine the maximal postirradiation time window for

Articles

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

Protocols

Protease Fluorescent Detection Kit detects protease activity using FITC-labeled casein substrate.

Protease Fluorescent Detection Kit detects protease activity using FITC-labeled casein substrate.

Protease Fluorescent Detection Kit detects protease activity using FITC-labeled casein substrate.

Protease Fluorescent Detection Kit detects protease activity using FITC-labeled casein substrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service