Skip to Content
Merck
All Photos(3)

Key Documents

P9703

Sigma-Aldrich

Pimaricin

from Streptomyces chattanoogensis, ≥95% (HPLC)

Synonym(s):

Natamycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H47NO13
CAS Number:
Molecular Weight:
665.73
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

biological source

Streptomyces chattanoogensis

Quality Level

Assay

≥95% (HPLC)

form

lyophilized powder
powder

solubility

0.1% formic acid in methanol: soluble 1 mg/mL

antibiotic activity spectrum

fungi

application(s)

agriculture
environmental

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

C[C@@H]1C\C=C\C=C\C=C\C=C\[C@@H](C[C@@H]2O[C@](O)(C[C@@H](O)C[C@H]3O[C@@H]3\C=C\C(=O)O1)C[C@H](O)[C@H]2C(O)=O)O[C@@H]4O[C@H](C)[C@@H](O)[C@H](N)[C@@H]4O

InChI

1S/C33H47NO13/c1-18-10-8-6-4-3-5-7-9-11-21(45-32-30(39)28(34)29(38)19(2)44-32)15-25-27(31(40)41)22(36)17-33(42,47-25)16-20(35)14-24-23(46-24)12-13-26(37)43-18/h3-9,11-13,18-25,27-30,32,35-36,38-39,42H,10,14-17,34H2,1-2H3,(H,40,41)/b4-3+,7-5+,8-6+,11-9+,13-12+/t18-,19-,20+,21+,22+,23-,24-,25+,27-,28+,29-,30+,32+,33-/m1/s1

InChI key

NCXMLFZGDNKEPB-FFPOYIOWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Pimaricin is an amphoteric antibiotic from Streptomyces chattanoogensis. It is used for a variety of fungal infections, mainly topically. It is used as an ergosterol and cholesterol binding agent to study lipid bilayer dynamics, especially in fungal cells. Pimaricin is used as a fungicide in agar media. It is used to study the pleitropic regulator AdpAch [1] and the gamma-butyrolactone regulatory system of Streptomyces chattanoogensis[2].

Biochem/physiol Actions

An antifungal polyene macrolide that binds specifically to ergosterol and blocks fungal growth. However, unlike nysatin and filipin, pimaricin does not change the permeability of the plasma membrane

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8 °C. Light sensitive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Ling Du et al.
Microbiology (Reading, England), 157(Pt 5), 1300-1311 (2011-02-19)
The complete natamycin (NTM) biosynthetic gene cluster of Streptomyces chattanoogensis was cloned and confirmed by the disruption of pathway-specific activator genes. Comparative cluster analysis with its counterpart in Streptomyces natalensis revealed different cluster architecture between these two clusters. Compared with
Gamma-Butyrolactone Regulatory System of Streptomyces chattanoogensis Links Nutrient Utilization, Metabolism, and Development
Yi-Ling Du, Shan-Zhen Li, et al.
Microbiology, 77, 8415-8426 (2011)
F N Arroyo-López et al.
International journal of food microbiology, 155(3), 257-262 (2012-03-01)
The present work uses a logistic/probabilistic model to obtain the growth/no growth interfaces of Saccharomyces cerevisiae, Wickerhamomyces anomalus and Candida boidinii (three yeast species commonly isolated from table olives) as a function of the diverse combinations of natamycin (0-30 mg/L)
Prajna Lalitha et al.
Cornea, 31(6), 662-667 (2012-02-16)
To analyze the minimum inhibitory concentration (MIC) of isolates from fungal keratitis to natamycin and voriconazole and to assess the relationship between organism, MIC, and clinical outcome. Data were collected as part of a randomized, controlled, double-masked clinical trial. Main
R S Bhatta et al.
International journal of pharmaceutics, 432(1-2), 105-112 (2012-05-10)
The aim of this study was to prepare natamycin encapsulated lecithin/chitosan mucoadhesive nanoparticles (NPs) for prolonged ocular application. These NPs were characterized by their mean particle size 213nm, encapsulation efficiency 73.57%, with a theoretical drug loading 5.09% and zeta potential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service