Skip to Content
Merck
All Photos(5)

Key Documents

P9129

Sigma-Aldrich

5-Pregnen-3β-ol-20-one

≥98%

Synonym(s):

3β-Hydroxy-5-pregnen-20-one, Pregnenolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H32O2
CAS Number:
Molecular Weight:
316.48
Beilstein:
2059026
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98%

form

powder

solubility

ethanol: soluble 10 mg/mL, clear, colorless to faintly yellow

functional group

ketone

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CC=C3C[C@@H](O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])C(C)=O

InChI

1S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,15-19,23H,5-12H2,1-3H3/t15-,16-,17+,18-,19-,20-,21+/m0/s1

InChI key

ORNBQBCIOKFOEO-QGVNFLHTSA-N

Gene Information

human ... SERPINA6(866)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

5-Pregnen-3β-ol-20-one is suitable for use:
  • as a substrate for 3β-HSD at a concentration of 1 μg/mL to study the time-dependent effect of reduced oxygen tension on 3β-hydroxysteroid dehydrogenase (3β-HSD) activity
  • as a progestin precursor to study the effects of splenic macrophages on progestin secretion of luteal cells
  • in the testicular interstitial cell culture medium at a concentration of 15mg/mL

Biochem/physiol Actions

One of a small number of "neurosteroids," steroids synthesized in the brain rather than peripheral sources. Decreased levels of these neurosteroids has been implicated in rodent cognitive decline with age, but studies in humans have been contradictory.
Testosterone production in rats occurs in Leydig cells of the testis via the Δ4 pathway. Cholesterol is first converted to pregnenolone, then to progesterone, and finally to testosterone. Pregnenolone is the key neurosteroid synthesized in steroidogenic glands. It is also present as a sulfate ester that serves as an antagonist of GABAergic neurons by interacting with γ-aminobutyric acid (GABA) receptor.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Matsuyama et al.
Endocrinologia japonica, 39(1), 51-57 (1992-02-01)
We have reported that splenic macrophages play a role in the regulation of progestin secretion in rats. In this study, splenic macrophages were obtained from cycling rats at different estrous cycle stages and co-cultured with luteal cells from mid-pseudopregnant rats
Jui-Hsia Weng et al.
Nature chemical biology, 9(10), 636-642 (2013-08-21)
Pregnenolone (P5) is a neurosteroid that improves memory and neurological recovery. It is also required for zebrafish embryonic development. However, its mode of action is unclear. Here we show that P5 promotes cell migration and microtubule polymerization by binding a
Z Y Hu et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(23), 8215-8219 (1987-12-01)
Oligodendrocyte mitochondria from 21-day-old Sprague-Dawley male rats were incubated with 100 nM [3H]cholesterol. It yielded [3H]pregnenolone at a rate of 2.5 +/- 0.7 and 5-[3H]pregnene-3 beta, 20 alpha-diol at a rate of 2.5 +/- 1.1 pmol per mg of protein
Nalaka S Rannulu et al.
Journal of the American Society for Mass Spectrometry, 23(9), 1558-1568 (2012-06-27)
The analysis of several bifunctional neutral steroids, 5-α-pregnane diol (5-α-pregnane-3α-20βdiol), estradiol (3,17α-dihydroxy-1,3,5(10)-estratriene), progesterone (4-pregnene-3,20-dione), lupeol (3β-hydroxy-20(29)-lupene), pregnenolone (5-pregnen-3β-ol-20-one), and pregnenolone acetate (5-pregnen-3β-ol-20-one acetate) was accomplished by negative ion electrospray mass spectrometry (ESI-MS) employing adduct formation with various anions: fluoride, bicarbonate
Ying-Tzu Chang et al.
Scientific reports, 9(1), 11628-11628 (2019-08-14)
Multidrug resistance presents an obstacle in cancer treatment. Among numerous combative strategies, collateral sensitivity (CS) drugs have opened a new avenue to defeat cancer by exploiting selective toxicity against multidrug-resistant (MDR) cancer. In the present study, a clinically used synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service